Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028696.hg.1 |
TC06002284.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028696.hg.1 |
ENST00000320285.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028696.hg.1 |
ENST00000366908.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028696.hg.1 |
ENST00000366911.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028696.hg.1 |
ENST00000457520.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028696.hg.1 |
NM_020133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028695.hg.1 |
TC06002284.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028695.hg.1 |
ENST00000320285.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028695.hg.1 |
ENST00000366908.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028695.hg.1 |
ENST00000366911.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028695.hg.1 |
ENST00000457520.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028695.hg.1 |
NM_020133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
ENST00000320285.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
ENST00000320285.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
ENST00000320285.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
ENST00000320285.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
ENST00000320285.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
ENST00000320285.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
ENST00000320285.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
ENST00000320285.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
ENST00000320285.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
ENST00000320285.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
ENST00000320285.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
ENST00000320285.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
ENST00000320285.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
ENST00000320285.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
ENST00000366905.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
ENST00000366905.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
ENST00000366905.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
ENST00000366905.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
ENST00000366905.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
ENST00000366911.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
ENST00000366911.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
ENST00000366911.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
ENST00000366911.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
ENST00000366911.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
ENST00000366911.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
ENST00000366911.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
ENST00000366911.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
ENST00000366911.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
ENST00000366911.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
ENST00000366911.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
ENST00000366911.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
ENST00000366911.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
ENST00000436279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
ENST00000436279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
ENST00000436279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
ENST00000436279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
ENST00000436279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
ENST00000436279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
ENST00000437165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
ENST00000437165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
ENST00000437165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
ENST00000437165.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
NM_020133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
NM_020133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
NM_020133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
NM_020133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
NM_020133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
NM_020133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
NM_020133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
NM_020133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
NM_020133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
NM_020133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
NM_020133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
NM_020133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
NM_020133.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013794.hg.1 |
NM_020133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
ENST00000320285.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
ENST00000320285.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
ENST00000320285.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
ENST00000320285.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
ENST00000320285.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
ENST00000320285.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
ENST00000320285.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
ENST00000320285.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
ENST00000320285.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
ENST00000320285.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
ENST00000320285.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
ENST00000320285.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
ENST00000320285.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
ENST00000320285.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
ENST00000366905.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
ENST00000366905.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
ENST00000366905.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
ENST00000366905.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
ENST00000366905.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
ENST00000366911.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
ENST00000366911.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
ENST00000366911.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
ENST00000366911.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
ENST00000366911.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
ENST00000366911.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
ENST00000366911.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
ENST00000366911.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
ENST00000366911.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
ENST00000366911.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
ENST00000366911.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
ENST00000366911.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
ENST00000366911.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
ENST00000436279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
ENST00000436279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
ENST00000436279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
ENST00000436279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
ENST00000436279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
ENST00000436279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
ENST00000437165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
ENST00000437165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
ENST00000437165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
ENST00000437165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
NM_020133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
NM_020133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
NM_020133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
NM_020133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
NM_020133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
NM_020133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
NM_020133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
NM_020133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
NM_020133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
NM_020133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
NM_020133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
NM_020133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
NM_020133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013794.hg.1 |
NM_020133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600195183.hg.1 |
ENST00000320285.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600195183.hg.1 |
ENST00000366911.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600195183.hg.1 |
ENST00000437165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600195183.hg.1 |
NM_020133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600195183.hg.1 |
ENST00000320285.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600195183.hg.1 |
ENST00000366911.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600195183.hg.1 |
ENST00000437165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600195183.hg.1 |
NM_020133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600195183.hg.1 |
TC0600013794.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600195182.hg.1 |
ENST00000320285.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600195182.hg.1 |
ENST00000366911.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600195182.hg.1 |
ENST00000436279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600195182.hg.1 |
ENST00000437165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600195182.hg.1 |
NM_020133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600195182.hg.1 |
ENST00000320285.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600195182.hg.1 |
ENST00000366911.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600195182.hg.1 |
ENST00000436279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600195182.hg.1 |
ENST00000437165.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600195182.hg.1 |
NM_020133.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600195182.hg.1 |
TC0600013794.hg.1 |
Clariom D TC |