Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000433878.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000433878.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000433878.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000433878.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000433878.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000433878.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000433878.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000433878.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000433878.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000433878.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000438446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000438446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000438446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000438446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000438446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000438446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000438446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000438446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000438446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000438446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000471739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000471739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000471739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000471739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000471739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010933.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03023095.hg.1 |
TC03001353.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03023095.hg.1 |
ENST00000296137.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03023095.hg.1 |
ENST00000438446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03023095.hg.1 |
ENST00000535325.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03023095.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300179682.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300179682.hg.1 |
ENST00000433878.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300179682.hg.1 |
ENST00000438446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300179682.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300179682.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300179682.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300179682.hg.1 |
ENST00000433878.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300179682.hg.1 |
ENST00000438446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300179682.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300179682.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300179682.hg.1 |
TC0300010933.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000296137.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000433878.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000433878.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000433878.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000433878.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000433878.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000433878.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000433878.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000433878.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000433878.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000433878.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000438446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000438446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000438446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000438446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000438446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000438446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000438446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000438446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000438446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000438446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000471739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000471739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000471739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000471739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000471739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
ENST00000535325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
NM_024513.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010933.hg.1 |
NM_024513.3 |
Refseq |