Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
ENST00000265132.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
ENST00000265132.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
ENST00000265132.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
ENST00000265132.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
ENST00000265132.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
ENST00000265132.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
ENST00000265132.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
ENST00000265132.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
ENST00000265132.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
ENST00000265132.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
ENST00000265132.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
ENST00000265132.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
ENST00000265132.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
ENST00000265132.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
ENST00000265132.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
ENST00000265132.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
ENST00000265132.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
ENST00000466610.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
ENST00000466610.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
ENST00000466610.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
ENST00000466610.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
ENST00000466610.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
ENST00000466610.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
ENST00000466610.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
ENST00000540645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
ENST00000540645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
ENST00000540645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
ENST00000540645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
ENST00000540645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
ENST00000540645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
ENST00000540645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
ENST00000540645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
ENST00000540645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
ENST00000540645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
ENST00000540645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
ENST00000540645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
ENST00000540645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
ENST00000603230.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
ENST00000603230.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
ENST00000603230.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
ENST00000603230.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
ENST00000603230.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
ENST00000603230.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
ENST00000603230.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
ENST00000603230.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
ENST00000603230.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
ENST00000603230.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
NM_001633.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
NM_001633.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
NM_001633.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
NM_001633.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
NM_001633.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
NM_001633.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
NM_001633.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
NM_001633.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
NM_001633.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
NM_001633.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
NM_001633.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
NM_001633.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
NM_001633.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
NM_001633.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
NM_001633.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
NM_001633.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011276.hg.1 |
NM_001633.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09018809.hg.1 |
TC09001506.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09018809.hg.1 |
ENST00000265132.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09018809.hg.1 |
ENST00000466610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09018809.hg.1 |
ENST00000540645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09018809.hg.1 |
NM_001633.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09018811.hg.1 |
TC09001506.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09018811.hg.1 |
ENST00000265132.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09018811.hg.1 |
ENST00000466610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09018811.hg.1 |
ENST00000540645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09018811.hg.1 |
NM_001633.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09018812.hg.1 |
TC09001506.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09018812.hg.1 |
ENST00000265132.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09018812.hg.1 |
ENST00000466610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09018812.hg.1 |
ENST00000540645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09018812.hg.1 |
NM_001633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900176850.hg.1 |
ENST00000265132.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176850.hg.1 |
ENST00000466610.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176850.hg.1 |
ENST00000540645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176850.hg.1 |
ENST00000603230.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176850.hg.1 |
NM_001633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900176850.hg.1 |
ENST00000265132.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176850.hg.1 |
ENST00000466610.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176850.hg.1 |
ENST00000540645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176850.hg.1 |
ENST00000603230.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176850.hg.1 |
NM_001633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900176850.hg.1 |
TC0900011276.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
ENST00000265132.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
ENST00000265132.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
ENST00000265132.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
ENST00000265132.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
ENST00000265132.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
ENST00000265132.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
ENST00000265132.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
ENST00000265132.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
ENST00000265132.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
ENST00000265132.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
ENST00000265132.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
ENST00000265132.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
ENST00000265132.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
ENST00000265132.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
ENST00000265132.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
ENST00000265132.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
ENST00000265132.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
ENST00000466610.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
ENST00000466610.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
ENST00000466610.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
ENST00000466610.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
ENST00000466610.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
ENST00000466610.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
ENST00000466610.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
ENST00000540645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
ENST00000540645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
ENST00000540645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
ENST00000540645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
ENST00000540645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
ENST00000540645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
ENST00000540645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
ENST00000540645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
ENST00000540645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
ENST00000540645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
ENST00000540645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
ENST00000540645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
ENST00000540645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
ENST00000603230.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
ENST00000603230.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
ENST00000603230.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
ENST00000603230.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
ENST00000603230.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
ENST00000603230.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
ENST00000603230.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
ENST00000603230.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
ENST00000603230.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
ENST00000603230.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
NM_001633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
NM_001633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
NM_001633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
NM_001633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
NM_001633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
NM_001633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
NM_001633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
NM_001633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
NM_001633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
NM_001633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
NM_001633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
NM_001633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
NM_001633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
NM_001633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
NM_001633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
NM_001633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011276.hg.1 |
NM_001633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900176852.hg.1 |
ENST00000265132.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176852.hg.1 |
ENST00000466610.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176852.hg.1 |
ENST00000540645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176852.hg.1 |
ENST00000603230.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176852.hg.1 |
NM_001633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900176852.hg.1 |
ENST00000265132.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176852.hg.1 |
ENST00000466610.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176852.hg.1 |
ENST00000540645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176852.hg.1 |
ENST00000603230.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176852.hg.1 |
NM_001633.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900176852.hg.1 |
TC0900011276.hg.1 |
Clariom D TC |