Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY327404.1 | - | 10 - 11 | 1315 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |