Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022771.4 | NP_073608.4 | 7 - 8 | 777 | 81 | |
XM_006719564.2 | XP_006719627.1 | 7 - 8 | 759 | 81 | ||
XM_006719565.3 | XP_006719628.1 | 7 - 8 | 697 | 81 | ||
XM_006719566.2 | XP_006719629.1 | 8 - 9 | 908 | 81 | ||
XM_011538679.1 | XP_011536981.1 | 9 - 10 | 834 | 81 | ||
XM_011538680.1 | XP_011536982.1 | 7 - 8 | 792 | 81 | ||
XM_011538681.2 | XP_011536983.1 | 7 - 8 | 741 | 81 | ||
XM_017019824.1 | XP_016875313.1 | 8 - 9 | 794 | 81 | ||
XM_017019827.1 | XP_016875316.1 | 4 - 5 | 417 | 81 | ||
GenBank mRNA | BC028352.2 | - | 7 - 8 | 725 | 4827246 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |