Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD300LF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289083.1 | NP_001276012.1 | 5 - 6 | 739 | 100 | |
NM_001289084.1 | NP_001276013.1 | 4 - 5 | 738 | 100 | ||
NM_001289085.1 | NP_001276014.1 | 5 - 6 | 867 | 100 | ||
NM_139018.4 | NP_620587.2 | 4 - 5 | 693 | 100 | ||
XM_017024212.1 | XP_016879701.1 | 4 - 5 | 802 | 100 | ||
XM_017024213.1 | XP_016879702.1 | 5 - 6 | 864 | 100 | ||
XM_017024214.1 | XP_016879703.1 | 5 - 6 | 854 | 100 | ||
XM_017024215.1 | XP_016879704.1 | 5 - 6 | 748 | 100 | ||
GenBank mRNA | AF251706.1 | - | 4 - 5 | 565 | 100 | |
AF375480.1 | - | 4 - 5 | 623 | 100 | ||
AK092757.1 | - | 5 - 6 | 693 | 100 | ||
AK298752.1 | - | 4 - 5 | 679 | 100 | ||
AK315165.1 | - | 4 - 5 | 616 | 100 | ||
AY303545.1 | - | 4 - 5 | 583 | 100 | ||
AY358545.1 | - | 4 - 5 | 640 | 100 | ||
BI911496.1 | - | 4 - 5 | 589 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |