Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022996.hg.1 |
ENST00000270238.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022996.hg.1 |
NM_001080434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022996.hg.1 |
TC19001684.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022997.hg.1 |
ENST00000270238.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022997.hg.1 |
NM_001080434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022997.hg.1 |
TC19001684.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022994.hg.1 |
ENST00000270238.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022994.hg.1 |
NM_001080434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022994.hg.1 |
TC19001684.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022995.hg.1 |
ENST00000270238.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022995.hg.1 |
NM_001080434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19022995.hg.1 |
TC19001684.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011077.hg.1 |
ENST00000270238.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011077.hg.1 |
ENST00000270238.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011077.hg.1 |
ENST00000270238.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011077.hg.1 |
ENST00000270238.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011077.hg.1 |
ENST00000270238.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011077.hg.1 |
ENST00000270238.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011077.hg.1 |
ENST00000270238.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011077.hg.1 |
ENST00000270238.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011077.hg.1 |
ENST00000270238.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011077.hg.1 |
ENST00000270238.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011077.hg.1 |
ENST00000270238.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011077.hg.1 |
ENST00000270238.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011077.hg.1 |
ENST00000270238.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011077.hg.1 |
ENST00000270238.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011077.hg.1 |
ENST00000270238.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011077.hg.1 |
ENST00000270238.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011077.hg.1 |
ENST00000600059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011077.hg.1 |
ENST00000600059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011077.hg.1 |
ENST00000600059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011077.hg.1 |
ENST00000600059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011077.hg.1 |
ENST00000600059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011077.hg.1 |
ENST00000600059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011077.hg.1 |
ENST00000600059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011077.hg.1 |
ENST00000600059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011077.hg.1 |
ENST00000600059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011077.hg.1 |
ENST00000600059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011077.hg.1 |
ENST00000600059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011077.hg.1 |
ENST00000600059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011077.hg.1 |
ENST00000600059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011077.hg.1 |
ENST00000600059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011077.hg.1 |
ENST00000600059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011077.hg.1 |
ENST00000600059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011077.hg.1 |
NM_001080434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011077.hg.1 |
NM_001080434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011077.hg.1 |
NM_001080434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011077.hg.1 |
NM_001080434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011077.hg.1 |
NM_001080434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011077.hg.1 |
NM_001080434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011077.hg.1 |
NM_001080434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011077.hg.1 |
NM_001080434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011077.hg.1 |
NM_001080434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011077.hg.1 |
NM_001080434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011077.hg.1 |
NM_001080434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011077.hg.1 |
NM_001080434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011077.hg.1 |
NM_001080434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011077.hg.1 |
NM_001080434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011077.hg.1 |
NM_001080434.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011077.hg.1 |
NM_001080434.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900192571.hg.1 |
ENST00000270238.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900192571.hg.1 |
ENST00000600059.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900192571.hg.1 |
NM_001080434.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900192571.hg.1 |
TC1900011077.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900192571.hg.1 |
ENST00000270238.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900192571.hg.1 |
ENST00000600059.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900192571.hg.1 |
NM_001080434.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900192569.hg.1 |
ENST00000270238.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900192569.hg.1 |
ENST00000600059.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900192569.hg.1 |
NM_001080434.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900192569.hg.1 |
TC1900011077.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900192569.hg.1 |
ENST00000270238.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900192569.hg.1 |
ENST00000600059.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900192569.hg.1 |
NM_001080434.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011077.hg.1 |
ENST00000270238.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011077.hg.1 |
ENST00000270238.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011077.hg.1 |
ENST00000270238.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011077.hg.1 |
ENST00000270238.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011077.hg.1 |
ENST00000270238.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011077.hg.1 |
ENST00000270238.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011077.hg.1 |
ENST00000270238.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011077.hg.1 |
ENST00000270238.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011077.hg.1 |
ENST00000270238.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011077.hg.1 |
ENST00000270238.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011077.hg.1 |
ENST00000270238.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011077.hg.1 |
ENST00000270238.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011077.hg.1 |
ENST00000270238.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011077.hg.1 |
ENST00000270238.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011077.hg.1 |
ENST00000270238.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011077.hg.1 |
ENST00000270238.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011077.hg.1 |
ENST00000600059.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011077.hg.1 |
ENST00000600059.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011077.hg.1 |
ENST00000600059.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011077.hg.1 |
ENST00000600059.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011077.hg.1 |
ENST00000600059.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011077.hg.1 |
ENST00000600059.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011077.hg.1 |
ENST00000600059.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011077.hg.1 |
ENST00000600059.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011077.hg.1 |
ENST00000600059.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011077.hg.1 |
ENST00000600059.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011077.hg.1 |
ENST00000600059.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011077.hg.1 |
ENST00000600059.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011077.hg.1 |
ENST00000600059.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011077.hg.1 |
ENST00000600059.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011077.hg.1 |
ENST00000600059.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011077.hg.1 |
ENST00000600059.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011077.hg.1 |
NM_001080434.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011077.hg.1 |
NM_001080434.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011077.hg.1 |
NM_001080434.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011077.hg.1 |
NM_001080434.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011077.hg.1 |
NM_001080434.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011077.hg.1 |
NM_001080434.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011077.hg.1 |
NM_001080434.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011077.hg.1 |
NM_001080434.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011077.hg.1 |
NM_001080434.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011077.hg.1 |
NM_001080434.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011077.hg.1 |
NM_001080434.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011077.hg.1 |
NM_001080434.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011077.hg.1 |
NM_001080434.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011077.hg.1 |
NM_001080434.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011077.hg.1 |
NM_001080434.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011077.hg.1 |
NM_001080434.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900192573.hg.1 |
ENST00000270238.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900192573.hg.1 |
NM_001080434.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900192573.hg.1 |
TC1900011077.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900192573.hg.1 |
ENST00000270238.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900192573.hg.1 |
NM_001080434.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900192570.hg.1 |
ENST00000270238.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900192570.hg.1 |
ENST00000600059.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900192570.hg.1 |
NM_001080434.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900192570.hg.1 |
TC1900011077.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900192570.hg.1 |
ENST00000270238.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900192570.hg.1 |
ENST00000600059.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900192570.hg.1 |
NM_001080434.1 |
Refseq |