Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKAP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005751.4 | NP_005742.4 | 13 - 14 | 4176 | 149 | |
NM_147185.2 | NP_671714.1 | 13 - 14 | 4176 | 149 | ||
XM_017011642.1 | XP_016867131.1 | 13 - 14 | 4184 | 149 | ||
XM_017011643.1 | XP_016867132.1 | 13 - 14 | 4184 | 149 | ||
XM_017011644.1 | XP_016867133.1 | 13 - 14 | 4184 | 149 | ||
XM_017011645.1 | XP_016867134.1 | 12 - 13 | 4130 | 149 | ||
XM_017011646.1 | XP_016867135.1 | 13 - 14 | 4184 | 149 | ||
XM_017011647.1 | XP_016867136.1 | 13 - 14 | 4184 | 149 | ||
XM_017011648.1 | XP_016867137.1 | 13 - 14 | 4184 | 149 | ||
XM_017011649.1 | XP_016867138.1 | 13 - 14 | 4184 | 149 | ||
XM_017011650.1 | XP_016867139.1 | 13 - 14 | 4184 | 149 | ||
XM_017011651.1 | XP_016867140.1 | 13 - 14 | 4184 | 149 | ||
XM_017011652.1 | XP_016867141.1 | 13 - 14 | 4184 | 149 | ||
XM_017011653.1 | XP_016867142.1 | 13 - 14 | 4184 | 149 | ||
XM_017011654.1 | XP_016867143.1 | 13 - 14 | 4184 | 149 | ||
XM_017011655.1 | XP_016867144.1 | 13 - 14 | 3843 | 149 | ||
XM_017011656.1 | XP_016867145.1 | 12 - 13 | 3773 | 149 | ||
GenBank mRNA | AB019691.1 | - | 13 - 14 | 4037 | 149 | |
AF026245.1 | - | 14 - 15 | 4171 | 149 | ||
AF083037.1 | - | 6 - 7 | 3021 | 149 | ||
AF247727.4 | - | 13 - 14 | 3951 | 149 | ||
AJ131693.1 | - | 13 - 14 | 4173 | 149 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons |