Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026345.hg.1 |
ENST00000335209.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026345.hg.1 |
ENST00000392545.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026345.hg.1 |
NM_017899.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026345.hg.1 |
NR_031766.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12026345.hg.1 |
TC12002012.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000335209.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000335209.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000335209.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000335209.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000335209.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000335209.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000335209.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000335209.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000335209.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000335209.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000335209.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000462502.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000462502.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000462502.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000462502.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000462502.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000462502.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000462502.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000462502.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000462502.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000462502.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000470612.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000470612.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000470612.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000470612.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000470612.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000470612.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000470612.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000470612.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000470612.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000470612.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000470612.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000482176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000482176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000482176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000482176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000482176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000482176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000482176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000482176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000535198.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000535198.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000535198.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000535198.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000541210.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000541210.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000541210.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000541210.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000541210.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000541210.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000541210.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000541210.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000541210.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000541210.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000549210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000549210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000549210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000549210.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000552139.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
ENST00000552139.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
NM_001168325.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
NM_001168325.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
NM_001168325.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
NM_001168325.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
NM_001168325.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
NM_001168325.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
NM_001168325.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
NM_001168325.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
NM_001168325.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
NM_001168325.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
NM_017899.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
NM_017899.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
NM_017899.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
NM_017899.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
NM_017899.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
NM_017899.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
NM_017899.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
NM_017899.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
NM_017899.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
NM_017899.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
NM_017899.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
NR_031766.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
NR_031766.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
NR_031766.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
NR_031766.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
NR_031766.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
NR_031766.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
NR_031766.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
NR_031766.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
NR_031766.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
NR_031766.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012080.hg.1 |
NR_031766.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200193963.hg.1 |
ENST00000335209.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200193963.hg.1 |
ENST00000462502.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200193963.hg.1 |
ENST00000470612.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200193963.hg.1 |
ENST00000482176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200193963.hg.1 |
NM_017899.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200193963.hg.1 |
NR_031766.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200193963.hg.1 |
ENST00000335209.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200193963.hg.1 |
ENST00000462502.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200193963.hg.1 |
ENST00000470612.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200193963.hg.1 |
ENST00000482176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200193963.hg.1 |
NM_017899.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200193963.hg.1 |
NR_031766.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200193963.hg.1 |
TC1200012080.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000335209.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000335209.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000335209.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000335209.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000335209.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000335209.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000335209.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000335209.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000335209.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000335209.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000335209.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000462502.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000462502.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000462502.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000462502.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000462502.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000462502.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000462502.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000462502.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000462502.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000462502.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000470612.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000470612.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000470612.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000470612.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000470612.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000470612.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000470612.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000470612.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000470612.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000470612.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000470612.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000482176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000482176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000482176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000482176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000482176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000482176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000482176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000482176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000535198.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000535198.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000535198.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000535198.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000541210.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000541210.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000541210.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000541210.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000541210.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000541210.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000541210.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000541210.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000541210.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000541210.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000549210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000549210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000549210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000549210.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000552139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
ENST00000552139.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
NM_001168325.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
NM_001168325.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
NM_001168325.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
NM_001168325.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
NM_001168325.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
NM_001168325.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
NM_001168325.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
NM_001168325.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
NM_001168325.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
NM_001168325.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
NM_017899.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
NM_017899.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
NM_017899.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
NM_017899.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
NM_017899.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
NM_017899.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
NM_017899.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
NM_017899.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
NM_017899.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
NM_017899.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
NM_017899.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
NR_031766.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
NR_031766.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
NR_031766.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
NR_031766.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
NR_031766.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
NR_031766.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
NR_031766.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
NR_031766.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
NR_031766.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
NR_031766.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012080.hg.1 |
NR_031766.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200193964.hg.1 |
ENST00000335209.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200193964.hg.1 |
ENST00000462502.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200193964.hg.1 |
ENST00000470612.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200193964.hg.1 |
ENST00000482176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200193964.hg.1 |
ENST00000541210.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200193964.hg.1 |
NM_001168325.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200193964.hg.1 |
NM_017899.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200193964.hg.1 |
NR_031766.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200193964.hg.1 |
ENST00000335209.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200193964.hg.1 |
ENST00000462502.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200193964.hg.1 |
ENST00000470612.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200193964.hg.1 |
ENST00000482176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200193964.hg.1 |
ENST00000541210.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200193964.hg.1 |
NM_001168325.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200193964.hg.1 |
NM_017899.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200193964.hg.1 |
NR_031766.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200193964.hg.1 |
TC1200012080.hg.1 |
Clariom D TC |