Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003968.3 | NP_003959.3 | 6 - 7 | 451 | 139 | |
NM_198195.1 | NP_937838.1 | 5 - 6 | 409 | 139 | ||
XM_011534210.1 | XP_011532512.1 | 5 - 6 | 365 | 139 | ||
XM_011534211.1 | XP_011532513.1 | 4 - 5 | 318 | 139 | ||
GenBank mRNA | AB012190.1 | - | 4 - 5 | 368 | 139 | |
AF046024.1 | - | 5 - 6 | 452 | 139 | ||
AK002159.1 | - | 5 - 6 | 396 | 139 | ||
AK289392.1 | - | 6 - 7 | 437 | 139 | ||
AK314751.1 | - | 6 - 7 | 434 | 139 | ||
BC022853.1 | - | 6 - 7 | 450 | 4073511 | 139 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |