Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02025909.hg.1 |
TC02001616.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02025909.hg.1 |
ENST00000175091.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02025909.hg.1 |
NM_014713.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011889.hg.1 |
ENST00000175091.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011889.hg.1 |
ENST00000175091.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011889.hg.1 |
ENST00000175091.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011889.hg.1 |
ENST00000175091.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011889.hg.1 |
ENST00000175091.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011889.hg.1 |
ENST00000175091.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011889.hg.1 |
ENST00000175091.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011889.hg.1 |
ENST00000175091.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011889.hg.1 |
ENST00000175091.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011889.hg.1 |
ENST00000175091.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011889.hg.1 |
ENST00000175091.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011889.hg.1 |
ENST00000175091.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011889.hg.1 |
ENST00000483117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011889.hg.1 |
ENST00000483117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011889.hg.1 |
ENST00000483117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011889.hg.1 |
ENST00000483117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011889.hg.1 |
ENST00000483117.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011889.hg.1 |
NM_014713.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011889.hg.1 |
NM_014713.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011889.hg.1 |
NM_014713.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011889.hg.1 |
NM_014713.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011889.hg.1 |
NM_014713.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011889.hg.1 |
NM_014713.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011889.hg.1 |
NM_014713.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011889.hg.1 |
NM_014713.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011889.hg.1 |
NM_014713.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011889.hg.1 |
NM_014713.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011889.hg.1 |
NM_014713.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200011889.hg.1 |
NM_014713.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011889.hg.1 |
ENST00000175091.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011889.hg.1 |
ENST00000175091.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011889.hg.1 |
ENST00000175091.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011889.hg.1 |
ENST00000175091.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011889.hg.1 |
ENST00000175091.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011889.hg.1 |
ENST00000175091.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011889.hg.1 |
ENST00000175091.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011889.hg.1 |
ENST00000175091.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011889.hg.1 |
ENST00000175091.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011889.hg.1 |
ENST00000175091.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011889.hg.1 |
ENST00000175091.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011889.hg.1 |
ENST00000175091.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011889.hg.1 |
ENST00000483117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011889.hg.1 |
ENST00000483117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011889.hg.1 |
ENST00000483117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011889.hg.1 |
ENST00000483117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011889.hg.1 |
ENST00000483117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200011889.hg.1 |
NM_014713.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011889.hg.1 |
NM_014713.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011889.hg.1 |
NM_014713.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011889.hg.1 |
NM_014713.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011889.hg.1 |
NM_014713.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011889.hg.1 |
NM_014713.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011889.hg.1 |
NM_014713.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011889.hg.1 |
NM_014713.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011889.hg.1 |
NM_014713.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011889.hg.1 |
NM_014713.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011889.hg.1 |
NM_014713.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200011889.hg.1 |
NM_014713.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200186364.hg.1 |
ENST00000175091.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200186364.hg.1 |
ENST00000483117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200186364.hg.1 |
NM_014713.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200186364.hg.1 |
ENST00000175091.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200186364.hg.1 |
ENST00000483117.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200186364.hg.1 |
NM_014713.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200186364.hg.1 |
TC0200011889.hg.1 |
Clariom D TC |