Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR18000098.hg.1 |
ENST00000261597.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18000098.hg.1 |
ENST00000543946.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18000098.hg.1 |
NM_006101.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18000098.hg.1 |
TC18000009.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006484.hg.1 |
ENST00000261597.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006484.hg.1 |
ENST00000261597.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006484.hg.1 |
ENST00000261597.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006484.hg.1 |
ENST00000261597.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006484.hg.1 |
ENST00000261597.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006484.hg.1 |
ENST00000261597.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006484.hg.1 |
ENST00000261597.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006484.hg.1 |
ENST00000261597.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006484.hg.1 |
ENST00000261597.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006484.hg.1 |
ENST00000261597.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006484.hg.1 |
ENST00000261597.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006484.hg.1 |
ENST00000261597.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006484.hg.1 |
ENST00000261597.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006484.hg.1 |
ENST00000261597.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006484.hg.1 |
ENST00000261597.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006484.hg.1 |
ENST00000261597.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006484.hg.1 |
ENST00000261597.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006484.hg.1 |
ENST00000261597.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006484.hg.1 |
ENST00000261597.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006484.hg.1 |
ENST00000574096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006484.hg.1 |
ENST00000574096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006484.hg.1 |
ENST00000574567.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006484.hg.1 |
ENST00000574567.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006484.hg.1 |
ENST00000575515.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006484.hg.1 |
ENST00000575515.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006484.hg.1 |
ENST00000575515.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006484.hg.1 |
ENST00000575515.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006484.hg.1 |
ENST00000575515.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006484.hg.1 |
ENST00000576274.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006484.hg.1 |
ENST00000576274.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006484.hg.1 |
ENST00000576274.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006484.hg.1 |
ENST00000576274.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006484.hg.1 |
NM_006101.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006484.hg.1 |
NM_006101.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006484.hg.1 |
NM_006101.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006484.hg.1 |
NM_006101.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006484.hg.1 |
NM_006101.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006484.hg.1 |
NM_006101.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006484.hg.1 |
NM_006101.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006484.hg.1 |
NM_006101.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006484.hg.1 |
NM_006101.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006484.hg.1 |
NM_006101.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006484.hg.1 |
NM_006101.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006484.hg.1 |
NM_006101.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006484.hg.1 |
NM_006101.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006484.hg.1 |
NM_006101.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006484.hg.1 |
NM_006101.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006484.hg.1 |
NM_006101.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006484.hg.1 |
NM_006101.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006484.hg.1 |
NM_006101.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800006484.hg.1 |
NM_006101.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800144393.hg.1 |
ENST00000261597.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800144393.hg.1 |
NM_006101.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800144393.hg.1 |
ENST00000261597.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800144393.hg.1 |
NM_006101.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800144393.hg.1 |
TC1800006484.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1800006484.hg.1 |
ENST00000261597.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006484.hg.1 |
ENST00000261597.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006484.hg.1 |
ENST00000261597.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006484.hg.1 |
ENST00000261597.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006484.hg.1 |
ENST00000261597.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006484.hg.1 |
ENST00000261597.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006484.hg.1 |
ENST00000261597.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006484.hg.1 |
ENST00000261597.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006484.hg.1 |
ENST00000261597.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006484.hg.1 |
ENST00000261597.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006484.hg.1 |
ENST00000261597.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006484.hg.1 |
ENST00000261597.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006484.hg.1 |
ENST00000261597.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006484.hg.1 |
ENST00000261597.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006484.hg.1 |
ENST00000261597.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006484.hg.1 |
ENST00000261597.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006484.hg.1 |
ENST00000261597.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006484.hg.1 |
ENST00000261597.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006484.hg.1 |
ENST00000261597.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006484.hg.1 |
ENST00000574096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006484.hg.1 |
ENST00000574096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006484.hg.1 |
ENST00000574567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006484.hg.1 |
ENST00000574567.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006484.hg.1 |
ENST00000575515.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006484.hg.1 |
ENST00000575515.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006484.hg.1 |
ENST00000575515.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006484.hg.1 |
ENST00000575515.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006484.hg.1 |
ENST00000575515.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006484.hg.1 |
ENST00000576274.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006484.hg.1 |
ENST00000576274.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006484.hg.1 |
ENST00000576274.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006484.hg.1 |
ENST00000576274.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800006484.hg.1 |
NM_006101.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006484.hg.1 |
NM_006101.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006484.hg.1 |
NM_006101.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006484.hg.1 |
NM_006101.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006484.hg.1 |
NM_006101.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006484.hg.1 |
NM_006101.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006484.hg.1 |
NM_006101.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006484.hg.1 |
NM_006101.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006484.hg.1 |
NM_006101.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006484.hg.1 |
NM_006101.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006484.hg.1 |
NM_006101.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006484.hg.1 |
NM_006101.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006484.hg.1 |
NM_006101.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006484.hg.1 |
NM_006101.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006484.hg.1 |
NM_006101.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006484.hg.1 |
NM_006101.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006484.hg.1 |
NM_006101.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006484.hg.1 |
NM_006101.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800006484.hg.1 |
NM_006101.2 |
Refseq |