Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01042248.hg.1 |
TC01002694.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01042248.hg.1 |
ENST00000371269.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01042248.hg.1 |
ENST00000535035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01042248.hg.1 |
ENST00000537443.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01042248.hg.1 |
ENST00000539536.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01042248.hg.1 |
NM_014762.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
ENST00000371269.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
ENST00000371269.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
ENST00000371269.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
ENST00000371269.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
ENST00000371269.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
ENST00000371269.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
ENST00000371269.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
ENST00000371269.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
ENST00000371269.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
ENST00000371269.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
ENST00000371269.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
ENST00000371269.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
ENST00000371269.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
ENST00000371269.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
ENST00000371269.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
ENST00000371269.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
ENST00000371269.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
ENST00000371269.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
ENST00000371269.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
ENST00000371269.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
ENST00000436604.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
ENST00000436604.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
ENST00000436604.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
ENST00000436604.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
ENST00000436604.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
ENST00000535035.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
ENST00000535035.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
ENST00000535035.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
ENST00000535035.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
ENST00000535035.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
ENST00000535035.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
ENST00000535035.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
ENST00000535035.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
ENST00000535035.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
ENST00000535035.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
ENST00000535035.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
ENST00000535035.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
ENST00000535035.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
ENST00000535035.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
ENST00000535035.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
ENST00000535035.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
ENST00000535035.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
ENST00000535035.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
NM_014762.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
NM_014762.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
NM_014762.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
NM_014762.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
NM_014762.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
NM_014762.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
NM_014762.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
NM_014762.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
NM_014762.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
NM_014762.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
NM_014762.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
NM_014762.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
NM_014762.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
NM_014762.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
NM_014762.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
NM_014762.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
NM_014762.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
NM_014762.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
NM_014762.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
NM_014762.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014276.hg.1 |
NM_014762.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100206988.hg.1 |
ENST00000371269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100206988.hg.1 |
ENST00000436604.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100206988.hg.1 |
ENST00000535035.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100206988.hg.1 |
NM_014762.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100206988.hg.1 |
ENST00000371269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100206988.hg.1 |
ENST00000436604.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100206988.hg.1 |
ENST00000535035.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100206988.hg.1 |
NM_014762.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100206988.hg.1 |
TC0100014276.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
ENST00000371269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
ENST00000371269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
ENST00000371269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
ENST00000371269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
ENST00000371269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
ENST00000371269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
ENST00000371269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
ENST00000371269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
ENST00000371269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
ENST00000371269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
ENST00000371269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
ENST00000371269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
ENST00000371269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
ENST00000371269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
ENST00000371269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
ENST00000371269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
ENST00000371269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
ENST00000371269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
ENST00000371269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
ENST00000371269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
ENST00000436604.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
ENST00000436604.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
ENST00000436604.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
ENST00000436604.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
ENST00000436604.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
ENST00000535035.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
ENST00000535035.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
ENST00000535035.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
ENST00000535035.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
ENST00000535035.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
ENST00000535035.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
ENST00000535035.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
ENST00000535035.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
ENST00000535035.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
ENST00000535035.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
ENST00000535035.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
ENST00000535035.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
ENST00000535035.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
ENST00000535035.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
ENST00000535035.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
ENST00000535035.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
ENST00000535035.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
ENST00000535035.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
NM_014762.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
NM_014762.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
NM_014762.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
NM_014762.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
NM_014762.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
NM_014762.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
NM_014762.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
NM_014762.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
NM_014762.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
NM_014762.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
NM_014762.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
NM_014762.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
NM_014762.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
NM_014762.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
NM_014762.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
NM_014762.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
NM_014762.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
NM_014762.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
NM_014762.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
NM_014762.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014276.hg.1 |
NM_014762.3 |
Refseq |