Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001158261.1 | NP_001151733.1 | 3 - 4 | 525 | 61 | |
NM_144681.2 | NP_653282.2 | 3 - 4 | 525 | 61 | ||
GenBank mRNA | AK057296.1 | - | 3 - 4 | 525 | 61 | |
BC029224.1 | - | 3 - 4 | 476 | 5172741 | 61 | |
CV030241.1 | - | 3 - 4 | 298 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons |