Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05028129.hg.1 |
TC05002041.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05028129.hg.1 |
ENST00000239231.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05028129.hg.1 |
ENST00000520504.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05028129.hg.1 |
NM_024594.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013399.hg.1 |
ENST00000520504.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013399.hg.1 |
ENST00000520504.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013399.hg.1 |
ENST00000520504.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013399.hg.1 |
ENST00000522176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013399.hg.1 |
ENST00000522176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013399.hg.1 |
ENST00000522176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013399.hg.1 |
ENST00000522176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013399.hg.1 |
ENST00000522176.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013399.hg.1 |
NM_024594.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013399.hg.1 |
NM_024594.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013399.hg.1 |
NM_024594.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013399.hg.1 |
NM_024594.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013399.hg.1 |
NM_024594.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013399.hg.1 |
NM_024594.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013399.hg.1 |
NM_024594.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013399.hg.1 |
NM_024594.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013399.hg.1 |
NM_024594.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013399.hg.1 |
NM_024594.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013399.hg.1 |
NM_024594.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013399.hg.1 |
NM_024594.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013399.hg.1 |
NM_024594.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013399.hg.1 |
NM_024594.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013399.hg.1 |
ENST00000520504.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013399.hg.1 |
ENST00000520504.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013399.hg.1 |
ENST00000520504.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013399.hg.1 |
ENST00000522176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013399.hg.1 |
ENST00000522176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013399.hg.1 |
ENST00000522176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013399.hg.1 |
ENST00000522176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013399.hg.1 |
ENST00000522176.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013399.hg.1 |
NM_024594.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013399.hg.1 |
NM_024594.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013399.hg.1 |
NM_024594.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013399.hg.1 |
NM_024594.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013399.hg.1 |
NM_024594.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013399.hg.1 |
NM_024594.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013399.hg.1 |
NM_024594.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013399.hg.1 |
NM_024594.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013399.hg.1 |
NM_024594.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013399.hg.1 |
NM_024594.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013399.hg.1 |
NM_024594.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013399.hg.1 |
NM_024594.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013399.hg.1 |
NM_024594.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013399.hg.1 |
NM_024594.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500197408.hg.1 |
ENST00000520504.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500197408.hg.1 |
NM_024594.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500197408.hg.1 |
ENST00000520504.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500197408.hg.1 |
NM_024594.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500197408.hg.1 |
TC0500013399.hg.1 |
Clariom D TC |