Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
ENST00000292432.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
ENST00000292432.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
ENST00000292432.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
ENST00000292432.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
ENST00000292432.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
ENST00000292432.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
ENST00000292432.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
ENST00000292432.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
ENST00000292432.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
ENST00000292432.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
ENST00000292432.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
ENST00000292432.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
ENST00000292432.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
ENST00000292432.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
ENST00000292432.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
ENST00000292432.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
ENST00000292432.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
ENST00000292432.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
ENST00000292432.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
ENST00000292432.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
ENST00000292432.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
ENST00000292432.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
ENST00000504910.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
ENST00000504910.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
ENST00000504910.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
ENST00000504910.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
ENST00000504910.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
ENST00000506834.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
ENST00000506834.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
ENST00000506834.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
ENST00000506834.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
ENST00000506834.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
ENST00000506834.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
ENST00000506834.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
ENST00000506834.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
ENST00000506834.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
ENST00000506834.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
ENST00000506834.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
ENST00000506834.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
ENST00000506834.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
ENST00000509717.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
ENST00000509717.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
ENST00000509717.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
ENST00000509717.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
ENST00000514058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
ENST00000514058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
ENST00000514058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
ENST00000514058.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
ENST00000514666.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
ENST00000514666.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
ENST00000514666.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
NM_002115.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
NM_002115.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
NM_002115.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
NM_002115.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
NM_002115.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
NM_002115.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
NM_002115.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
NM_002115.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
NM_002115.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
NM_002115.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
NM_002115.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
NM_002115.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
NM_002115.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
NM_002115.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
NM_002115.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
NM_002115.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
NM_002115.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
NM_002115.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
NM_002115.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
NM_002115.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
NM_002115.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012956.hg.1 |
NM_002115.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05028840.hg.1 |
TC05002100.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05028840.hg.1 |
ENST00000292432.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05028840.hg.1 |
NM_002115.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
ENST00000292432.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
ENST00000292432.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
ENST00000292432.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
ENST00000292432.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
ENST00000292432.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
ENST00000292432.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
ENST00000292432.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
ENST00000292432.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
ENST00000292432.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
ENST00000292432.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
ENST00000292432.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
ENST00000292432.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
ENST00000292432.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
ENST00000292432.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
ENST00000292432.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
ENST00000292432.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
ENST00000292432.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
ENST00000292432.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
ENST00000292432.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
ENST00000292432.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
ENST00000292432.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
ENST00000292432.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
ENST00000504910.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
ENST00000504910.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
ENST00000504910.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
ENST00000504910.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
ENST00000504910.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
ENST00000506834.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
ENST00000506834.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
ENST00000506834.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
ENST00000506834.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
ENST00000506834.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
ENST00000506834.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
ENST00000506834.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
ENST00000506834.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
ENST00000506834.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
ENST00000506834.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
ENST00000506834.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
ENST00000506834.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
ENST00000506834.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
ENST00000509717.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
ENST00000509717.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
ENST00000509717.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
ENST00000509717.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
ENST00000514058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
ENST00000514058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
ENST00000514058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
ENST00000514058.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
ENST00000514666.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
ENST00000514666.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
ENST00000514666.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
NM_002115.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
NM_002115.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
NM_002115.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
NM_002115.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
NM_002115.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
NM_002115.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
NM_002115.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
NM_002115.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
NM_002115.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
NM_002115.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
NM_002115.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
NM_002115.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
NM_002115.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
NM_002115.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
NM_002115.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
NM_002115.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
NM_002115.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
NM_002115.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
NM_002115.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
NM_002115.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
NM_002115.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012956.hg.1 |
NM_002115.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500190191.hg.1 |
ENST00000292432.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500190191.hg.1 |
ENST00000504910.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500190191.hg.1 |
NM_002115.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500190191.hg.1 |
ENST00000292432.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500190191.hg.1 |
ENST00000504910.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500190191.hg.1 |
NM_002115.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500190191.hg.1 |
TC0500012956.hg.1 |
Clariom D TC |