Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007277.hg.1 |
ENST00000219660.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007277.hg.1 |
ENST00000219660.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007277.hg.1 |
ENST00000219660.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007277.hg.1 |
ENST00000219660.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007277.hg.1 |
ENST00000219660.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007277.hg.1 |
ENST00000219660.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007277.hg.1 |
ENST00000219660.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007277.hg.1 |
ENST00000219660.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007277.hg.1 |
ENST00000566125.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007277.hg.1 |
ENST00000566125.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007277.hg.1 |
ENST00000566125.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007277.hg.1 |
ENST00000566125.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007277.hg.1 |
ENST00000566125.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007277.hg.1 |
ENST00000566125.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007277.hg.1 |
ENST00000566125.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007277.hg.1 |
NM_001169.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007277.hg.1 |
NM_001169.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007277.hg.1 |
NM_001169.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007277.hg.1 |
NM_001169.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007277.hg.1 |
NM_001169.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007277.hg.1 |
NM_001169.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600007277.hg.1 |
NM_001169.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600152436.hg.1 |
ENST00000219660.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600152436.hg.1 |
ENST00000566125.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600152436.hg.1 |
NM_001169.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600152436.hg.1 |
ENST00000219660.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600152436.hg.1 |
ENST00000566125.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600152436.hg.1 |
NM_001169.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600152436.hg.1 |
TC1600007277.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600152432.hg.1 |
ENST00000566125.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600152432.hg.1 |
ENST00000566125.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600152432.hg.1 |
TC1600007277.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600007277.hg.1 |
ENST00000219660.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007277.hg.1 |
ENST00000219660.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007277.hg.1 |
ENST00000219660.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007277.hg.1 |
ENST00000219660.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007277.hg.1 |
ENST00000219660.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007277.hg.1 |
ENST00000219660.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007277.hg.1 |
ENST00000219660.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007277.hg.1 |
ENST00000219660.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007277.hg.1 |
ENST00000566125.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007277.hg.1 |
ENST00000566125.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007277.hg.1 |
ENST00000566125.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007277.hg.1 |
ENST00000566125.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007277.hg.1 |
ENST00000566125.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007277.hg.1 |
ENST00000566125.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007277.hg.1 |
ENST00000566125.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600007277.hg.1 |
NM_001169.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007277.hg.1 |
NM_001169.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007277.hg.1 |
NM_001169.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007277.hg.1 |
NM_001169.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007277.hg.1 |
NM_001169.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007277.hg.1 |
NM_001169.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600007277.hg.1 |
NM_001169.2 |
Refseq |