Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM245 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032012.3 | NP_114401.2 | 7 - 8 | 1377 | 125 | |
XM_011518446.2 | XP_011516748.1 | 7 - 8 | 1392 | 125 | ||
XM_011518449.1 | XP_011516751.1 | 6 - 7 | 1260 | 125 | ||
XM_011518452.1 | XP_011516754.1 | 6 - 7 | 1143 | 125 | ||
GenBank mRNA | AF153415.1 | - | 7 - 8 | 1377 | 125 | |
AK299952.1 | - | 7 - 8 | 1359 | 125 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |