Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR21003920.hg.1 |
ENST00000284987.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21003920.hg.1 |
NM_007038.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21003920.hg.1 |
TC21000346.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007822.hg.1 |
ENST00000284987.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007822.hg.1 |
ENST00000284987.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007822.hg.1 |
ENST00000284987.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007822.hg.1 |
ENST00000284987.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007822.hg.1 |
ENST00000284987.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007822.hg.1 |
ENST00000284987.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007822.hg.1 |
ENST00000284987.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007822.hg.1 |
ENST00000284987.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007822.hg.1 |
ENST00000284987.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007822.hg.1 |
ENST00000284987.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007822.hg.1 |
ENST00000284987.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007822.hg.1 |
ENST00000284987.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007822.hg.1 |
ENST00000284987.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007822.hg.1 |
NM_007038.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007822.hg.1 |
NM_007038.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007822.hg.1 |
NM_007038.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007822.hg.1 |
NM_007038.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007822.hg.1 |
NM_007038.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007822.hg.1 |
NM_007038.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007822.hg.1 |
NM_007038.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007822.hg.1 |
NM_007038.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007822.hg.1 |
NM_007038.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007822.hg.1 |
NM_007038.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007822.hg.1 |
NM_007038.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007822.hg.1 |
NM_007038.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007822.hg.1 |
NM_007038.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100007822.hg.1 |
NM_007038.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007822.hg.1 |
ENST00000284987.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007822.hg.1 |
ENST00000284987.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007822.hg.1 |
ENST00000284987.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007822.hg.1 |
ENST00000284987.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007822.hg.1 |
ENST00000284987.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007822.hg.1 |
ENST00000284987.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007822.hg.1 |
ENST00000284987.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007822.hg.1 |
ENST00000284987.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007822.hg.1 |
ENST00000284987.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007822.hg.1 |
ENST00000284987.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007822.hg.1 |
ENST00000284987.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007822.hg.1 |
ENST00000284987.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007822.hg.1 |
ENST00000284987.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100007822.hg.1 |
NM_007038.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007822.hg.1 |
NM_007038.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007822.hg.1 |
NM_007038.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007822.hg.1 |
NM_007038.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007822.hg.1 |
NM_007038.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007822.hg.1 |
NM_007038.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007822.hg.1 |
NM_007038.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007822.hg.1 |
NM_007038.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007822.hg.1 |
NM_007038.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007822.hg.1 |
NM_007038.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007822.hg.1 |
NM_007038.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007822.hg.1 |
NM_007038.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007822.hg.1 |
NM_007038.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100007822.hg.1 |
NM_007038.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100152736.hg.1 |
ENST00000284987.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100152736.hg.1 |
NM_007038.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100152736.hg.1 |
TC2100007822.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100152736.hg.1 |
ENST00000284987.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100152736.hg.1 |
NM_007038.4 |
Refseq |