Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000474438.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000474438.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000474438.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000474438.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000474438.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000474438.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164846.hg.1 |
ENST00000372262.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164846.hg.1 |
ENST00000537909.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164846.hg.1 |
NM_021248.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000164846.hg.1 |
TC2000009263.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000164846.hg.1 |
ENST00000372262.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164846.hg.1 |
ENST00000537909.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000164846.hg.1 |
NM_021248.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000372262.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000474438.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000474438.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000474438.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000474438.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000474438.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000474438.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
ENST00000537909.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009263.hg.1 |
NM_021248.2 |
Refseq |