Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEMA6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300780.1 | NP_001287709.1 | 17 - 18 | 2502 | 140 | |
GenBank mRNA | AB037789.1 | - | 17 - 18 | 1971 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons |