Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06029305.hg.1 |
TC06002335.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06029305.hg.1 |
ENST00000339209.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06029305.hg.1 |
ENST00000366780.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06029305.hg.1 |
NM_018288.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06029305.hg.1 |
NM_133325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000339209.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000339209.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000339209.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000339209.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000339209.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000339209.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000339209.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000339209.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000339209.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000339209.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000339209.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000339209.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000339209.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000339209.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000339209.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000339209.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000366780.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000366780.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000366780.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000366780.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000366780.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000366780.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000366780.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000366780.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000366780.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000366780.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000366780.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000366780.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000366780.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000366780.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000464779.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000464779.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000464779.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000464779.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000464779.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000480008.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000480008.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000480008.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000480008.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000480008.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000480008.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000480008.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000480008.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000480008.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000480008.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000480008.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000612128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000612128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000612128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000612128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000612128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000612128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000612128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000612128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000612128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000612128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000612128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
ENST00000612128.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
NM_018288.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
NM_018288.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
NM_018288.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
NM_018288.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
NM_018288.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
NM_018288.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
NM_018288.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
NM_018288.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
NM_018288.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
NM_018288.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
NM_018288.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
NM_018288.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
NM_018288.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
NM_018288.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
NM_018288.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
NM_133325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
NM_133325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
NM_133325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
NM_133325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
NM_133325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
NM_133325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
NM_133325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
NM_133325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
NM_133325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
NM_133325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
NM_133325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
NM_133325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
NM_133325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
NM_133325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014374.hg.1 |
NM_133325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000339209.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000339209.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000339209.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000339209.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000339209.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000339209.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000339209.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000339209.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000339209.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000339209.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000339209.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000339209.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000339209.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000339209.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000339209.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000339209.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000366780.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000366780.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000366780.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000366780.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000366780.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000366780.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000366780.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000366780.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000366780.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000366780.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000366780.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000366780.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000366780.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000366780.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000464779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000464779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000464779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000464779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000464779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000480008.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000480008.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000480008.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000480008.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000480008.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000480008.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000480008.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000480008.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000480008.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000480008.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000480008.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000612128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000612128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000612128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000612128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000612128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000612128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000612128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000612128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000612128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000612128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000612128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
ENST00000612128.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
NM_018288.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
NM_018288.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
NM_018288.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
NM_018288.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
NM_018288.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
NM_018288.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
NM_018288.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
NM_018288.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
NM_018288.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
NM_018288.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
NM_018288.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
NM_018288.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
NM_018288.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
NM_018288.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
NM_018288.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
NM_133325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
NM_133325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
NM_133325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
NM_133325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
NM_133325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
NM_133325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
NM_133325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
NM_133325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
NM_133325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
NM_133325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
NM_133325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
NM_133325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
NM_133325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
NM_133325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014374.hg.1 |
NM_133325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600203249.hg.1 |
ENST00000339209.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600203249.hg.1 |
ENST00000366780.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600203249.hg.1 |
NM_018288.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600203249.hg.1 |
NM_133325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600203249.hg.1 |
ENST00000339209.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600203249.hg.1 |
ENST00000366780.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600203249.hg.1 |
NM_018288.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600203249.hg.1 |
NM_133325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600203249.hg.1 |
TC0600014374.hg.1 |
Clariom D TC |