Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR13001837.hg.1 |
ENST00000379221.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13001837.hg.1 |
ENST00000474320.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13001837.hg.1 |
NM_013238.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13001837.hg.1 |
TC13000157.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009954.hg.1 |
ENST00000379221.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009954.hg.1 |
ENST00000379221.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009954.hg.1 |
ENST00000379221.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009954.hg.1 |
ENST00000379221.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009954.hg.1 |
ENST00000379221.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009954.hg.1 |
ENST00000379221.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009954.hg.1 |
ENST00000379221.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009954.hg.1 |
ENST00000379221.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009954.hg.1 |
ENST00000379221.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009954.hg.1 |
ENST00000379221.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009954.hg.1 |
ENST00000379221.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009954.hg.1 |
ENST00000474320.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009954.hg.1 |
ENST00000474320.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009954.hg.1 |
ENST00000474320.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009954.hg.1 |
ENST00000474320.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009954.hg.1 |
ENST00000474320.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009954.hg.1 |
ENST00000474320.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009954.hg.1 |
ENST00000474320.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009954.hg.1 |
NM_013238.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009954.hg.1 |
NM_013238.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009954.hg.1 |
NM_013238.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009954.hg.1 |
NM_013238.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009954.hg.1 |
NM_013238.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009954.hg.1 |
NM_013238.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009954.hg.1 |
NM_013238.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009954.hg.1 |
NM_013238.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009954.hg.1 |
NM_013238.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009954.hg.1 |
NM_013238.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009954.hg.1 |
ENST00000379221.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009954.hg.1 |
ENST00000379221.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009954.hg.1 |
ENST00000379221.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009954.hg.1 |
ENST00000379221.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009954.hg.1 |
ENST00000379221.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009954.hg.1 |
ENST00000379221.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009954.hg.1 |
ENST00000379221.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009954.hg.1 |
ENST00000379221.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009954.hg.1 |
ENST00000379221.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009954.hg.1 |
ENST00000379221.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009954.hg.1 |
ENST00000379221.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009954.hg.1 |
ENST00000474320.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009954.hg.1 |
ENST00000474320.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009954.hg.1 |
ENST00000474320.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009954.hg.1 |
ENST00000474320.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009954.hg.1 |
ENST00000474320.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009954.hg.1 |
ENST00000474320.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009954.hg.1 |
ENST00000474320.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009954.hg.1 |
NM_013238.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009954.hg.1 |
NM_013238.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009954.hg.1 |
NM_013238.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009954.hg.1 |
NM_013238.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009954.hg.1 |
NM_013238.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009954.hg.1 |
NM_013238.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009954.hg.1 |
NM_013238.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009954.hg.1 |
NM_013238.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009954.hg.1 |
NM_013238.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009954.hg.1 |
NM_013238.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300166467.hg.1 |
ENST00000379221.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300166467.hg.1 |
ENST00000474320.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300166467.hg.1 |
NM_013238.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300166467.hg.1 |
ENST00000379221.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300166467.hg.1 |
ENST00000474320.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300166467.hg.1 |
NM_013238.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300166467.hg.1 |
TC1300009954.hg.1 |
Clariom D TC |