Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR2000170521.hg.1 |
ENST00000328969.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170521.hg.1 |
ENST00000355969.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170521.hg.1 |
ENST00000357119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170521.hg.1 |
ENST00000369967.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170521.hg.1 |
ENST00000448100.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170521.hg.1 |
ENST00000472711.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170521.hg.1 |
ENST00000477340.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170521.hg.1 |
ENST00000478385.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170521.hg.1 |
ENST00000484569.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170521.hg.1 |
ENST00000490623.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170521.hg.1 |
NM_001083113.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000170521.hg.1 |
NM_001195653.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000170521.hg.1 |
NM_001195654.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000170521.hg.1 |
NM_032527.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000170521.hg.1 |
NM_181485.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000170521.hg.1 |
TC2000009954.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000170521.hg.1 |
ENST00000328969.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170521.hg.1 |
ENST00000355969.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170521.hg.1 |
ENST00000357119.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170521.hg.1 |
ENST00000369967.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170521.hg.1 |
ENST00000448100.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170521.hg.1 |
ENST00000472711.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170521.hg.1 |
ENST00000477340.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170521.hg.1 |
ENST00000478385.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170521.hg.1 |
ENST00000484569.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170521.hg.1 |
ENST00000490623.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000170521.hg.1 |
NM_001083113.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000170521.hg.1 |
NM_001195653.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000170521.hg.1 |
NM_001195654.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000170521.hg.1 |
NM_032527.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000170521.hg.1 |
NM_181485.2 |
Refseq |