Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009418.hg.1 |
TC0X000732.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009418.hg.1 |
ENST00000370357.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009418.hg.1 |
NM_173493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
ENST00000370357.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
ENST00000370357.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
ENST00000370357.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
ENST00000370357.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
ENST00000370357.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
ENST00000370357.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
ENST00000370357.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
ENST00000370357.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
ENST00000370357.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
ENST00000370357.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
ENST00000370357.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
ENST00000370357.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
ENST00000370357.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
ENST00000370357.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
ENST00000370357.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
ENST00000370357.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
ENST00000370357.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
ENST00000370357.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
ENST00000370357.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
ENST00000370357.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
ENST00000413236.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
ENST00000413236.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
ENST00000413236.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
ENST00000413236.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
ENST00000413236.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
ENST00000413236.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
ENST00000464219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
ENST00000464219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
ENST00000464219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
ENST00000464219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
ENST00000464219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
ENST00000464219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
ENST00000464219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
ENST00000464219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
ENST00000464219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
ENST00000464219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
ENST00000464219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
ENST00000464219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
ENST00000464219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
ENST00000464219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
ENST00000464219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
ENST00000464219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
ENST00000464219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
ENST00000464219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
ENST00000464219.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
NM_173493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
NM_173493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
NM_173493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
NM_173493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
NM_173493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
NM_173493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
NM_173493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
NM_173493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
NM_173493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
NM_173493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
NM_173493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
NM_173493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
NM_173493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
NM_173493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
NM_173493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
NM_173493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
NM_173493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
NM_173493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
NM_173493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
NM_173493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008734.hg.1 |
NM_173493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
ENST00000370357.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
ENST00000370357.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
ENST00000370357.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
ENST00000370357.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
ENST00000370357.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
ENST00000370357.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
ENST00000370357.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
ENST00000370357.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
ENST00000370357.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
ENST00000370357.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
ENST00000370357.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
ENST00000370357.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
ENST00000370357.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
ENST00000370357.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
ENST00000370357.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
ENST00000370357.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
ENST00000370357.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
ENST00000370357.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
ENST00000370357.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
ENST00000370357.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
ENST00000413236.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
ENST00000413236.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
ENST00000413236.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
ENST00000413236.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
ENST00000413236.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
ENST00000413236.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
ENST00000464219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
ENST00000464219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
ENST00000464219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
ENST00000464219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
ENST00000464219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
ENST00000464219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
ENST00000464219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
ENST00000464219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
ENST00000464219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
ENST00000464219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
ENST00000464219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
ENST00000464219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
ENST00000464219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
ENST00000464219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
ENST00000464219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
ENST00000464219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
ENST00000464219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
ENST00000464219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
ENST00000464219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
NM_173493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
NM_173493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
NM_173493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
NM_173493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
NM_173493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
NM_173493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
NM_173493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
NM_173493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
NM_173493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
NM_173493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
NM_173493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
NM_173493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
NM_173493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
NM_173493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
NM_173493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
NM_173493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
NM_173493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
NM_173493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
NM_173493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
NM_173493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008734.hg.1 |
NM_173493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00159326.hg.1 |
ENST00000370357.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00159326.hg.1 |
ENST00000413236.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00159326.hg.1 |
ENST00000464219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00159326.hg.1 |
NM_173493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00159326.hg.1 |
ENST00000370357.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00159326.hg.1 |
ENST00000413236.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00159326.hg.1 |
ENST00000464219.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00159326.hg.1 |
NM_173493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00159326.hg.1 |
TC0X00008734.hg.1 |
Clariom D TC |