Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011540542.1 | XP_011538844.1 | 1 - 2 | 116 | 123 | |
XM_017000134.1 | XP_016855623.1 | 1 - 2 | 121 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |