Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFTPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005264487.2 | XP_005264544.1 | 10 - 11 | 1212 | 60 | |
GenBank mRNA | AB439566.1 | - | 1 - 2 | 55 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Homo sapiens Probe spans exons |