Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VASP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003370.3 | NP_003361.1 | 6 - 7 | 1063 | 81 | |
XM_005259200.1 | XP_005259257.1 | 6 - 7 | 1051 | 81 | ||
GenBank mRNA | AK314812.1 | - | 6 - 7 | 1030 | 81 | |
AL556269.3 | - | 4 - 5 | 536 | 81 | ||
BC038224.1 | - | 6 - 7 | 981 | 5210128 | 81 | |
BG825983.1 | - | 4 - 5 | 80 | |||
KU178381.1 | - | 5 - 6 | 721 | 81 | ||
KU178382.1 | - | 6 - 7 | 565 | 81 | ||
KU178385.1 | - | 3 - 4 | 270 | 81 | ||
Z46389.1 | - | 6 - 7 | 975 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |