Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BIRC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016252.3 | NP_057336.3 | 23 - 24 | 4884 | 91 | |
XM_005264450.4 | XP_005264507.2 | 23 - 24 | 5047 | 91 | ||
XM_017004557.1 | XP_016860046.1 | 23 - 24 | 5047 | 91 | ||
GenBank mRNA | AF265555.1 | - | 23 - 24 | 4666 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Homo sapiens Probe spans exons |