Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04007599.hg.1 |
TC04000515.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04007599.hg.1 |
ENST00000326581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04007599.hg.1 |
ENST00000514652.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04007599.hg.1 |
NM_032149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04007598.hg.1 |
TC04000515.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04007598.hg.1 |
ENST00000326581.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04007598.hg.1 |
ENST00000514652.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04007598.hg.1 |
NM_032149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
ENST00000326581.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
ENST00000326581.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
ENST00000326581.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
ENST00000326581.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
ENST00000326581.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
ENST00000326581.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
ENST00000326581.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
ENST00000326581.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
ENST00000326581.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
ENST00000326581.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
ENST00000326581.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
ENST00000326581.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
ENST00000326581.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
ENST00000477187.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
ENST00000477187.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
ENST00000477187.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
ENST00000477187.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
ENST00000477187.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
ENST00000477187.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
ENST00000477187.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
ENST00000477187.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
ENST00000477187.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
ENST00000477187.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
ENST00000477187.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
ENST00000477187.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
ENST00000477187.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
ENST00000477187.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
ENST00000503257.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
ENST00000503257.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
ENST00000503257.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
ENST00000503257.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
ENST00000514652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
ENST00000514652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
ENST00000514652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
ENST00000514652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
ENST00000514652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
ENST00000514652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
ENST00000514652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
ENST00000514652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
ENST00000514652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
ENST00000514652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
ENST00000514652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
NM_032149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
NM_032149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
NM_032149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
NM_032149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
NM_032149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
NM_032149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
NM_032149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
NM_032149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
NM_032149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
NM_032149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
NM_032149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
NM_032149.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008236.hg.1 |
NM_032149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
ENST00000326581.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
ENST00000326581.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
ENST00000326581.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
ENST00000326581.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
ENST00000326581.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
ENST00000326581.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
ENST00000326581.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
ENST00000326581.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
ENST00000326581.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
ENST00000326581.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
ENST00000326581.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
ENST00000326581.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
ENST00000326581.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
ENST00000477187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
ENST00000477187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
ENST00000477187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
ENST00000477187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
ENST00000477187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
ENST00000477187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
ENST00000477187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
ENST00000477187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
ENST00000477187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
ENST00000477187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
ENST00000477187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
ENST00000477187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
ENST00000477187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
ENST00000477187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
ENST00000503257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
ENST00000503257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
ENST00000503257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
ENST00000503257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
ENST00000514652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
ENST00000514652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
ENST00000514652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
ENST00000514652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
ENST00000514652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
ENST00000514652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
ENST00000514652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
ENST00000514652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
ENST00000514652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
ENST00000514652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
ENST00000514652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
NM_032149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
NM_032149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
NM_032149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
NM_032149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
NM_032149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
NM_032149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
NM_032149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
NM_032149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
NM_032149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
NM_032149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
NM_032149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
NM_032149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008236.hg.1 |
NM_032149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400156667.hg.1 |
ENST00000326581.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156667.hg.1 |
ENST00000477187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156667.hg.1 |
ENST00000514652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156667.hg.1 |
NM_032149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400156667.hg.1 |
ENST00000326581.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156667.hg.1 |
ENST00000477187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156667.hg.1 |
ENST00000514652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156667.hg.1 |
NM_032149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400156667.hg.1 |
TC0400008236.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400156668.hg.1 |
ENST00000326581.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156668.hg.1 |
ENST00000477187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156668.hg.1 |
ENST00000514652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156668.hg.1 |
NM_032149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400156668.hg.1 |
ENST00000326581.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156668.hg.1 |
ENST00000477187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156668.hg.1 |
ENST00000514652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400156668.hg.1 |
NM_032149.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400156668.hg.1 |
TC0400008236.hg.1 |
Clariom D TC |