Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16000033.hg.1 |
ENST00000293861.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16000033.hg.1 |
ENST00000383018.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16000033.hg.1 |
NM_024571.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16000033.hg.1 |
TC16000003.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000293861.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000293861.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000293861.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000293861.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000293861.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000293861.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000293861.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000293861.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000293861.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000293861.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000293861.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000293861.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000383018.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000383018.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000383018.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000383018.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000383018.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000383018.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000383018.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000383018.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000383018.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000383018.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000383018.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000397876.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000397876.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000397876.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000397876.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000397876.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000397876.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000397876.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000397876.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000397876.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000417493.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000417493.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000417493.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000417493.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000417493.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000417493.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000466183.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000466183.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000466183.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000466183.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000466183.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000466183.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000466183.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000466183.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000481947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000481947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000481947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000481947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000481947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000481947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000481947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000481947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000481947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000493672.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000493672.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000493672.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000493672.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
ENST00000493672.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
NM_024571.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
NM_024571.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
NM_024571.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
NM_024571.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
NM_024571.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
NM_024571.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
NM_024571.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
NM_024571.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
NM_024571.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
NM_024571.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006436.hg.1 |
NM_024571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000293861.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000293861.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000293861.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000293861.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000293861.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000293861.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000293861.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000293861.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000293861.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000293861.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000293861.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000293861.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000383018.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000383018.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000383018.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000383018.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000383018.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000383018.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000383018.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000383018.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000383018.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000383018.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000383018.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000397876.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000397876.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000397876.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000397876.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000397876.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000397876.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000397876.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000397876.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000397876.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000417493.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000417493.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000417493.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000417493.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000417493.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000417493.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000466183.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000466183.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000466183.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000466183.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000466183.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000466183.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000466183.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000466183.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000481947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000481947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000481947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000481947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000481947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000481947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000481947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000481947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000481947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000493672.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000493672.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000493672.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000493672.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
ENST00000493672.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
NM_024571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
NM_024571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
NM_024571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
NM_024571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
NM_024571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
NM_024571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
NM_024571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
NM_024571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
NM_024571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
NM_024571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006436.hg.1 |
NM_024571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600144090.hg.1 |
ENST00000293861.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144090.hg.1 |
ENST00000383018.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144090.hg.1 |
ENST00000397876.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144090.hg.1 |
ENST00000417493.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144090.hg.1 |
ENST00000466183.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144090.hg.1 |
ENST00000481947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144090.hg.1 |
NM_024571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600144090.hg.1 |
ENST00000293861.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144090.hg.1 |
ENST00000383018.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144090.hg.1 |
ENST00000397876.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144090.hg.1 |
ENST00000417493.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144090.hg.1 |
ENST00000466183.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144090.hg.1 |
ENST00000481947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600144090.hg.1 |
NM_024571.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600144090.hg.1 |
TC1600006436.hg.1 |
Clariom D TC |