Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05016654.hg.1 |
TC05001166.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05016654.hg.1 |
ENST00000304623.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05016654.hg.1 |
ENST00000359640.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05016654.hg.1 |
ENST00000458100.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05016654.hg.1 |
ENST00000495388.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05016654.hg.1 |
ENST00000503622.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05016654.hg.1 |
ENST00000511377.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05016654.hg.1 |
ENST00000538638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05016654.hg.1 |
NM_001332.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05016652.hg.1 |
TC05001166.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05016652.hg.1 |
ENST00000304623.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05016652.hg.1 |
ENST00000359640.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05016652.hg.1 |
ENST00000458100.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05016652.hg.1 |
ENST00000495388.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05016652.hg.1 |
ENST00000503622.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05016652.hg.1 |
ENST00000511377.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05016652.hg.1 |
ENST00000538638.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05016652.hg.1 |
NM_001332.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500170920.hg.1 |
ENST00000304623.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500170920.hg.1 |
ENST00000458100.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500170920.hg.1 |
ENST00000495388.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500170920.hg.1 |
ENST00000503622.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500170920.hg.1 |
ENST00000504499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500170920.hg.1 |
ENST00000511377.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500170920.hg.1 |
ENST00000513588.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500170920.hg.1 |
NM_001288715.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500170920.hg.1 |
NM_001288716.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500170920.hg.1 |
NM_001288717.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500170920.hg.1 |
NM_001332.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500170920.hg.1 |
NR_109988.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500170920.hg.1 |
ENST00000304623.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500170920.hg.1 |
ENST00000458100.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500170920.hg.1 |
ENST00000495388.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500170920.hg.1 |
ENST00000503622.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500170920.hg.1 |
ENST00000504499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500170920.hg.1 |
ENST00000511377.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500170920.hg.1 |
ENST00000513588.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500170920.hg.1 |
NM_001288715.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500170920.hg.1 |
NM_001288716.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500170920.hg.1 |
NM_001288717.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500170920.hg.1 |
NM_001332.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500170920.hg.1 |
NR_109988.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500170920.hg.1 |
TC0500010115.hg.1 |
Clariom D TC |