Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17012987.hg.1 |
ENST00000320345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17012987.hg.1 |
ENST00000350761.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17012987.hg.1 |
ENST00000397335.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17012987.hg.1 |
ENST00000406424.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17012987.hg.1 |
ENST00000421807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17012987.hg.1 |
ENST00000542125.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17012987.hg.1 |
NM_001135642.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17012987.hg.1 |
NM_016532.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17012987.hg.1 |
NM_130766.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17012987.hg.1 |
TC17000975.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17012989.hg.1 |
ENST00000320345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17012989.hg.1 |
ENST00000350761.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17012989.hg.1 |
ENST00000397335.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17012989.hg.1 |
ENST00000406424.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17012989.hg.1 |
ENST00000421807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17012989.hg.1 |
ENST00000542125.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17012989.hg.1 |
NM_001135642.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17012989.hg.1 |
NM_016532.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17012989.hg.1 |
NM_130766.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17012989.hg.1 |
TC17000975.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17012985.hg.1 |
ENST00000320345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17012985.hg.1 |
ENST00000350761.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17012985.hg.1 |
ENST00000397335.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17012985.hg.1 |
ENST00000406424.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17012985.hg.1 |
ENST00000421807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17012985.hg.1 |
ENST00000542125.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17012985.hg.1 |
NM_001135642.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17012985.hg.1 |
NM_016532.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17012985.hg.1 |
NM_130766.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17012985.hg.1 |
TC17000975.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17012990.hg.1 |
ENST00000320345.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17012990.hg.1 |
ENST00000350761.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17012990.hg.1 |
ENST00000397335.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17012990.hg.1 |
ENST00000406424.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17012990.hg.1 |
ENST00000421807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17012990.hg.1 |
ENST00000542125.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17012990.hg.1 |
NM_001135642.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17012990.hg.1 |
NM_016532.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17012990.hg.1 |
NM_130766.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17012990.hg.1 |
TC17000975.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700171659.hg.1 |
ENST00000320345.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171659.hg.1 |
ENST00000350761.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171659.hg.1 |
ENST00000406424.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171659.hg.1 |
ENST00000421807.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171659.hg.1 |
ENST00000449479.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171659.hg.1 |
ENST00000461552.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171659.hg.1 |
ENST00000495339.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171659.hg.1 |
ENST00000574561.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171659.hg.1 |
NM_001135642.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700171659.hg.1 |
NM_016532.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700171659.hg.1 |
NM_130766.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700171659.hg.1 |
ENST00000320345.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171659.hg.1 |
ENST00000350761.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171659.hg.1 |
ENST00000406424.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171659.hg.1 |
ENST00000421807.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171659.hg.1 |
ENST00000449479.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171659.hg.1 |
ENST00000461552.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171659.hg.1 |
ENST00000495339.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171659.hg.1 |
ENST00000574561.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171659.hg.1 |
NM_001135642.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700171659.hg.1 |
NM_016532.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700171659.hg.1 |
NM_130766.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700171659.hg.1 |
TC1700009377.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700171664.hg.1 |
ENST00000320345.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171664.hg.1 |
ENST00000350761.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171664.hg.1 |
ENST00000406424.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171664.hg.1 |
ENST00000421807.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171664.hg.1 |
ENST00000449479.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171664.hg.1 |
ENST00000477115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171664.hg.1 |
ENST00000481867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171664.hg.1 |
ENST00000495339.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171664.hg.1 |
ENST00000575172.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171664.hg.1 |
NM_001135642.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700171664.hg.1 |
NM_016532.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700171664.hg.1 |
NM_130766.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700171664.hg.1 |
ENST00000320345.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171664.hg.1 |
ENST00000350761.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171664.hg.1 |
ENST00000406424.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171664.hg.1 |
ENST00000421807.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171664.hg.1 |
ENST00000449479.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171664.hg.1 |
ENST00000477115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171664.hg.1 |
ENST00000481867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171664.hg.1 |
ENST00000495339.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171664.hg.1 |
ENST00000575172.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171664.hg.1 |
NM_001135642.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700171664.hg.1 |
NM_016532.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700171664.hg.1 |
NM_130766.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700171664.hg.1 |
TC1700009377.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700171661.hg.1 |
ENST00000320345.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171661.hg.1 |
ENST00000350761.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171661.hg.1 |
ENST00000406424.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171661.hg.1 |
ENST00000421807.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171661.hg.1 |
ENST00000449479.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171661.hg.1 |
ENST00000461552.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171661.hg.1 |
ENST00000477115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171661.hg.1 |
ENST00000481867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171661.hg.1 |
ENST00000495339.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171661.hg.1 |
ENST00000574561.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171661.hg.1 |
NM_001135642.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700171661.hg.1 |
NM_016532.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700171661.hg.1 |
NM_130766.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700171661.hg.1 |
ENST00000320345.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171661.hg.1 |
ENST00000350761.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171661.hg.1 |
ENST00000406424.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171661.hg.1 |
ENST00000421807.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171661.hg.1 |
ENST00000449479.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171661.hg.1 |
ENST00000461552.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171661.hg.1 |
ENST00000477115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171661.hg.1 |
ENST00000481867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171661.hg.1 |
ENST00000495339.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171661.hg.1 |
ENST00000574561.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171661.hg.1 |
NM_001135642.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700171661.hg.1 |
NM_016532.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700171661.hg.1 |
NM_130766.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700171661.hg.1 |
TC1700009377.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700171665.hg.1 |
ENST00000320345.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171665.hg.1 |
ENST00000350761.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171665.hg.1 |
ENST00000406424.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171665.hg.1 |
ENST00000421807.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171665.hg.1 |
ENST00000449479.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171665.hg.1 |
ENST00000477115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171665.hg.1 |
ENST00000481867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171665.hg.1 |
ENST00000575172.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171665.hg.1 |
NM_001135642.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700171665.hg.1 |
NM_016532.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700171665.hg.1 |
NM_130766.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700171665.hg.1 |
ENST00000320345.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171665.hg.1 |
ENST00000350761.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171665.hg.1 |
ENST00000406424.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171665.hg.1 |
ENST00000421807.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171665.hg.1 |
ENST00000449479.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171665.hg.1 |
ENST00000477115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171665.hg.1 |
ENST00000481867.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171665.hg.1 |
ENST00000575172.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700171665.hg.1 |
NM_001135642.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700171665.hg.1 |
NM_016532.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700171665.hg.1 |
NM_130766.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700171665.hg.1 |
TC1700009377.hg.1 |
Clariom D TC |