Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008556.hg.1 |
ENST00000307128.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008556.hg.1 |
ENST00000307128.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008556.hg.1 |
ENST00000307128.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008556.hg.1 |
ENST00000307128.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008556.hg.1 |
ENST00000307128.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008556.hg.1 |
ENST00000307128.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008556.hg.1 |
ENST00000307128.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008556.hg.1 |
ENST00000307128.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008556.hg.1 |
ENST00000307128.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008556.hg.1 |
ENST00000307128.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008556.hg.1 |
NM_016599.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008556.hg.1 |
NM_016599.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008556.hg.1 |
NM_016599.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008556.hg.1 |
NM_016599.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008556.hg.1 |
NM_016599.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008556.hg.1 |
NM_016599.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008556.hg.1 |
NM_016599.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008556.hg.1 |
NM_016599.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008556.hg.1 |
NM_016599.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008556.hg.1 |
NM_016599.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04008801.hg.1 |
TC04000610.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04008801.hg.1 |
ENST00000307128.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04008801.hg.1 |
NM_016599.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04008798.hg.1 |
TC04000610.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04008798.hg.1 |
ENST00000307128.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04008798.hg.1 |
NM_016599.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400158719.hg.1 |
ENST00000307128.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400158719.hg.1 |
NM_016599.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400158719.hg.1 |
ENST00000307128.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400158719.hg.1 |
NM_016599.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400158719.hg.1 |
TC0400008556.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400158715.hg.1 |
ENST00000307128.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400158715.hg.1 |
NM_016599.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400158715.hg.1 |
ENST00000307128.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400158715.hg.1 |
NM_016599.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400158715.hg.1 |
TC0400008556.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400158716.hg.1 |
ENST00000307128.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400158716.hg.1 |
NM_016599.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400158716.hg.1 |
ENST00000307128.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400158716.hg.1 |
NM_016599.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400158716.hg.1 |
TC0400008556.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400008556.hg.1 |
ENST00000307128.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008556.hg.1 |
ENST00000307128.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008556.hg.1 |
ENST00000307128.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008556.hg.1 |
ENST00000307128.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008556.hg.1 |
ENST00000307128.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008556.hg.1 |
ENST00000307128.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008556.hg.1 |
ENST00000307128.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008556.hg.1 |
ENST00000307128.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008556.hg.1 |
ENST00000307128.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008556.hg.1 |
ENST00000307128.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400008556.hg.1 |
NM_016599.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008556.hg.1 |
NM_016599.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008556.hg.1 |
NM_016599.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008556.hg.1 |
NM_016599.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008556.hg.1 |
NM_016599.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008556.hg.1 |
NM_016599.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008556.hg.1 |
NM_016599.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008556.hg.1 |
NM_016599.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008556.hg.1 |
NM_016599.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400008556.hg.1 |
NM_016599.4 |
Refseq |