Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003636.hg.1 |
TC09000343.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003636.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003638.hg.1 |
TC09000343.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003638.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003638.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003640.hg.1 |
TC09000343.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003640.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003640.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003637.hg.1 |
TC09000343.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003637.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003637.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003641.hg.1 |
TC09000343.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003641.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003641.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003642.hg.1 |
TC09000343.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003642.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09003642.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900151394.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151394.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900151394.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151394.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900151394.hg.1 |
TC0900007604.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900151389.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151389.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900151389.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151389.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900151389.hg.1 |
TC0900007604.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900151395.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151395.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900151395.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900151395.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900151395.hg.1 |
TC0900007604.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900151388.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900151388.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900151388.hg.1 |
TC0900007604.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
ENST00000346234.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007604.hg.1 |
NM_012383.4 |
Refseq |