Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FANCI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113378.1 | NP_001106849.1 | 32 - 33 | 3629 | 103 | |
NM_018193.2 | NP_060663.2 | 31 - 32 | 3449 | 103 | ||
XM_011521756.2 | XP_011520058.1 | 32 - 33 | 3653 | 103 | ||
XM_011521757.2 | XP_011520059.1 | 32 - 33 | 3649 | 103 | ||
XM_011521764.2 | XP_011520066.1 | 31 - 32 | 3473 | 103 | ||
XM_011521767.2 | XP_011520069.1 | 32 - 33 | 3623 | 103 | ||
GenBank mRNA | AB058697.1 | - | 18 - 19 | 3553 | 103 | |
AK001581.1 | - | 25 - 26 | 2835 | 103 | ||
AK027564.1 | - | 20 - 21 | 2336 | 103 | ||
BC021859.1 | - | 9 - 10 | 971 | 103 | ||
BC140769.1 | - | 32 - 33 | 3586 | 9021637 | 103 | |
BC144483.1 | - | 32 - 33 | 3586 | 9053010 | 103 | |
EF469766.1 | - | 31 - 32 | 3539 | 103 | ||
EF567077.1 | - | 31 - 32 | 3539 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |