Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011966.hg.1 |
ENST00000488231.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011966.hg.1 |
ENST00000488231.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12024874.hg.1 |
ENST00000240055.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12024874.hg.1 |
ENST00000551727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12024874.hg.1 |
NM_006166.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12024874.hg.1 |
TC12001906.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011755.hg.1 |
ENST00000240055.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011755.hg.1 |
ENST00000240055.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011755.hg.1 |
ENST00000240055.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011755.hg.1 |
ENST00000240055.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011755.hg.1 |
ENST00000240055.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011755.hg.1 |
ENST00000240055.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011755.hg.1 |
ENST00000240055.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011755.hg.1 |
ENST00000240055.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011755.hg.1 |
ENST00000240055.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011755.hg.1 |
ENST00000240055.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011755.hg.1 |
ENST00000240055.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011755.hg.1 |
ENST00000240055.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011755.hg.1 |
ENST00000240055.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011755.hg.1 |
ENST00000550189.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011755.hg.1 |
ENST00000550189.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011755.hg.1 |
ENST00000550189.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011755.hg.1 |
ENST00000550189.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011755.hg.1 |
ENST00000550189.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011755.hg.1 |
ENST00000550189.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011755.hg.1 |
ENST00000550881.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011755.hg.1 |
ENST00000550881.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011755.hg.1 |
ENST00000550881.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011755.hg.1 |
ENST00000550881.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011755.hg.1 |
ENST00000551446.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011755.hg.1 |
ENST00000551446.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011755.hg.1 |
ENST00000551446.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011755.hg.1 |
ENST00000551446.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011755.hg.1 |
ENST00000551446.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011755.hg.1 |
ENST00000551446.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011755.hg.1 |
ENST00000551727.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011755.hg.1 |
ENST00000551727.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011755.hg.1 |
ENST00000551727.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011755.hg.1 |
ENST00000551727.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011755.hg.1 |
ENST00000551727.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011755.hg.1 |
ENST00000551727.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011755.hg.1 |
ENST00000551727.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011755.hg.1 |
ENST00000551727.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011755.hg.1 |
ENST00000551727.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011755.hg.1 |
ENST00000551727.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011755.hg.1 |
NM_006166.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011755.hg.1 |
NM_006166.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011755.hg.1 |
NM_006166.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011755.hg.1 |
NM_006166.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011755.hg.1 |
NM_006166.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011755.hg.1 |
NM_006166.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011755.hg.1 |
NM_006166.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011755.hg.1 |
NM_006166.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011755.hg.1 |
NM_006166.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011755.hg.1 |
NM_006166.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011755.hg.1 |
NM_006166.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011755.hg.1 |
NM_006166.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200191173.hg.1 |
ENST00000240055.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200191173.hg.1 |
ENST00000550189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200191173.hg.1 |
ENST00000551446.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200191173.hg.1 |
ENST00000551727.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200191173.hg.1 |
NM_006166.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200191173.hg.1 |
ENST00000240055.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200191173.hg.1 |
ENST00000550189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200191173.hg.1 |
ENST00000551446.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200191173.hg.1 |
ENST00000551727.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200191173.hg.1 |
NM_006166.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200191173.hg.1 |
TC1200011755.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300011966.hg.1 |
ENST00000488231.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011966.hg.1 |
ENST00000488231.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011755.hg.1 |
ENST00000240055.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011755.hg.1 |
ENST00000240055.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011755.hg.1 |
ENST00000240055.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011755.hg.1 |
ENST00000240055.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011755.hg.1 |
ENST00000240055.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011755.hg.1 |
ENST00000240055.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011755.hg.1 |
ENST00000240055.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011755.hg.1 |
ENST00000240055.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011755.hg.1 |
ENST00000240055.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011755.hg.1 |
ENST00000240055.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011755.hg.1 |
ENST00000240055.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011755.hg.1 |
ENST00000240055.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011755.hg.1 |
ENST00000240055.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011755.hg.1 |
ENST00000550189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011755.hg.1 |
ENST00000550189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011755.hg.1 |
ENST00000550189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011755.hg.1 |
ENST00000550189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011755.hg.1 |
ENST00000550189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011755.hg.1 |
ENST00000550189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011755.hg.1 |
ENST00000550881.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011755.hg.1 |
ENST00000550881.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011755.hg.1 |
ENST00000550881.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011755.hg.1 |
ENST00000550881.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011755.hg.1 |
ENST00000551446.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011755.hg.1 |
ENST00000551446.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011755.hg.1 |
ENST00000551446.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011755.hg.1 |
ENST00000551446.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011755.hg.1 |
ENST00000551446.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011755.hg.1 |
ENST00000551446.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011755.hg.1 |
ENST00000551727.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011755.hg.1 |
ENST00000551727.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011755.hg.1 |
ENST00000551727.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011755.hg.1 |
ENST00000551727.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011755.hg.1 |
ENST00000551727.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011755.hg.1 |
ENST00000551727.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011755.hg.1 |
ENST00000551727.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011755.hg.1 |
ENST00000551727.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011755.hg.1 |
ENST00000551727.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011755.hg.1 |
ENST00000551727.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011755.hg.1 |
NM_006166.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011755.hg.1 |
NM_006166.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011755.hg.1 |
NM_006166.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011755.hg.1 |
NM_006166.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011755.hg.1 |
NM_006166.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011755.hg.1 |
NM_006166.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011755.hg.1 |
NM_006166.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011755.hg.1 |
NM_006166.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011755.hg.1 |
NM_006166.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011755.hg.1 |
NM_006166.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011755.hg.1 |
NM_006166.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011755.hg.1 |
NM_006166.3 |
Refseq |