Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17020082.hg.1 |
ENST00000310778.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17020082.hg.1 |
ENST00000341193.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17020082.hg.1 |
ENST00000347901.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17020082.hg.1 |
ENST00000393939.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17020082.hg.1 |
NM_001079870.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17020082.hg.1 |
NM_001079871.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17020082.hg.1 |
NM_177977.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17020082.hg.1 |
TC17001516.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700184604.hg.1 |
ENST00000310778.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700184604.hg.1 |
ENST00000341193.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700184604.hg.1 |
ENST00000347901.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700184604.hg.1 |
ENST00000393939.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700184604.hg.1 |
NM_001079870.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700184604.hg.1 |
NM_001079871.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700184604.hg.1 |
NM_177977.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700184604.hg.1 |
ENST00000310778.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700184604.hg.1 |
ENST00000341193.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700184604.hg.1 |
ENST00000347901.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700184604.hg.1 |
ENST00000393939.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700184604.hg.1 |
NM_001079870.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700184604.hg.1 |
NM_001079871.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700184604.hg.1 |
NM_177977.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700184604.hg.1 |
TC1700010684.hg.1 |
Clariom D TC |