Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYOM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152372.3 | NP_689585.3 | 35 - 36 | 4160 | 65 | |
GenBank mRNA | AF086254.1 | - | 4 - 5 | 250 | 65 | |
AK093280.1 | - | 8 - 9 | 1046 | 65 | ||
AK128233.1 | - | 27 - 28 | 3220 | 65 | ||
BC014671.1 | - | 9 - 10 | 680 | 65 | ||
BX537911.1 | - | 34 - 35 | 3884 | 65 | ||
BX647453.1 | - | 35 - 36 | 4121 | 65 | ||
BX647455.1 | - | 35 - 36 | 4160 | 65 | ||
BX647644.1 | - | 35 - 36 | 4121 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons |