Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DTNBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271667.1 | NP_001258596.1 | 6 - 7 | 727 | 87 | |
NM_001271668.1 | NP_001258597.1 | 5 - 6 | 631 | 87 | ||
NM_001271669.1 | NP_001258598.1 | 4 - 5 | 577 | 87 | ||
NM_032122.4 | NP_115498.2 | 6 - 7 | 682 | 87 | ||
NM_183040.2 | NP_898861.1 | 6 - 7 | 682 | 87 | ||
NR_036448.1 | 7 - 8 | 821 | 87 | |||
XM_005249447.4 | XP_005249504.1 | 7 - 8 | 683 | 87 | ||
XM_011514936.2 | XP_011513238.1 | 6 - 7 | 673 | 87 | ||
GenBank mRNA | AF061734.1 | - | 2 - 3 | 169 | 87 | |
AF394226.1 | - | 6 - 7 | 599 | 87 | ||
AK054593.1 | - | 6 - 7 | 651 | 87 | ||
AK290718.1 | - | 6 - 7 | 599 | 87 | ||
AK310590.1 | - | 7 - 8 | 683 | 87 | ||
AL136637.2 | - | 6 - 7 | 599 | 87 | ||
AY265460.1 | - | 6 - 7 | 606 | 87 | ||
BC011912.2 | - | 6 - 7 | 597 | 4139934 | 87 | |
BE793298.1 | - | 539 | 87 | |||
BQ056746.1 | - | 5 - 6 | 570 | 87 | ||
BX394617.2 | - | 7 - 8 | 821 | 87 | ||
KU178801.1 | - | 6 - 7 | 493 | 87 | ||
KU178802.1 | - | 4 - 5 | 388 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons |