Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRGAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020762.2 | NP_065813.1 | 14 - 15 | 1735 | 144 | |
XM_005269042.3 | XP_005269099.1 | 14 - 15 | 2055 | 144 | ||
XM_011538580.1 | XP_011536882.1 | 15 - 16 | 1843 | 144 | ||
XM_011538581.2 | XP_011536883.1 | 15 - 16 | 3348 | 144 | ||
GenBank mRNA | AB037725.1 | - | 13 - 14 | 2957 | 144 | |
AK023899.1 | - | 4 - 5 | 465 | 144 | ||
BC053903.1 | - | 14 - 15 | 1735 | 6526787 | 144 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Homo sapiens Probe spans exons |