Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010708.hg.1 |
ENST00000263368.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010708.hg.1 |
ENST00000263368.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010708.hg.1 |
ENST00000263368.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010708.hg.1 |
ENST00000263368.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010708.hg.1 |
ENST00000263368.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010708.hg.1 |
ENST00000263368.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010708.hg.1 |
ENST00000263368.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010708.hg.1 |
ENST00000263368.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010708.hg.1 |
ENST00000263368.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010708.hg.1 |
ENST00000595483.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010708.hg.1 |
ENST00000595483.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010708.hg.1 |
ENST00000595483.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010708.hg.1 |
ENST00000595483.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010708.hg.1 |
ENST00000595483.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010708.hg.1 |
ENST00000595483.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010708.hg.1 |
ENST00000595483.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010708.hg.1 |
ENST00000597870.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010708.hg.1 |
ENST00000597870.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010708.hg.1 |
ENST00000597870.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010708.hg.1 |
ENST00000597870.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010708.hg.1 |
ENST00000597870.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010708.hg.1 |
ENST00000597870.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010708.hg.1 |
ENST00000597870.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010708.hg.1 |
ENST00000597870.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010708.hg.1 |
ENST00000597870.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010708.hg.1 |
ENST00000601346.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010708.hg.1 |
ENST00000601346.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010708.hg.1 |
ENST00000601346.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010708.hg.1 |
ENST00000601346.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010708.hg.1 |
ENST00000601346.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010708.hg.1 |
ENST00000601346.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010708.hg.1 |
NM_000713.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010708.hg.1 |
NM_000713.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010708.hg.1 |
NM_000713.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010708.hg.1 |
NM_000713.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010708.hg.1 |
NM_000713.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010708.hg.1 |
NM_000713.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010708.hg.1 |
NM_000713.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010708.hg.1 |
NM_000713.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010708.hg.1 |
NM_000713.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19021021.hg.1 |
ENST00000263368.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19021021.hg.1 |
NM_000713.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19021021.hg.1 |
TC19001542.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19021022.hg.1 |
ENST00000263368.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19021022.hg.1 |
NM_000713.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19021022.hg.1 |
TC19001542.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900188514.hg.1 |
ENST00000263368.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900188514.hg.1 |
ENST00000595483.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900188514.hg.1 |
ENST00000597870.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900188514.hg.1 |
ENST00000601346.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900188514.hg.1 |
NM_000713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900188514.hg.1 |
TC1900010708.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900188514.hg.1 |
ENST00000263368.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900188514.hg.1 |
ENST00000595483.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900188514.hg.1 |
ENST00000597870.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900188514.hg.1 |
ENST00000601346.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900188514.hg.1 |
NM_000713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900188511.hg.1 |
ENST00000263368.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900188511.hg.1 |
ENST00000597870.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900188511.hg.1 |
ENST00000601346.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900188511.hg.1 |
NM_000713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900188511.hg.1 |
TC1900010708.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900188511.hg.1 |
ENST00000263368.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900188511.hg.1 |
ENST00000597870.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900188511.hg.1 |
ENST00000601346.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900188511.hg.1 |
NM_000713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900188516.hg.1 |
ENST00000263368.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900188516.hg.1 |
ENST00000595483.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900188516.hg.1 |
ENST00000597870.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900188516.hg.1 |
ENST00000601346.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900188516.hg.1 |
NM_000713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900188516.hg.1 |
TC1900010708.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900188516.hg.1 |
ENST00000263368.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900188516.hg.1 |
ENST00000595483.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900188516.hg.1 |
ENST00000597870.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900188516.hg.1 |
ENST00000601346.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900188516.hg.1 |
NM_000713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010708.hg.1 |
ENST00000263368.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010708.hg.1 |
ENST00000263368.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010708.hg.1 |
ENST00000263368.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010708.hg.1 |
ENST00000263368.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010708.hg.1 |
ENST00000263368.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010708.hg.1 |
ENST00000263368.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010708.hg.1 |
ENST00000263368.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010708.hg.1 |
ENST00000263368.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010708.hg.1 |
ENST00000263368.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010708.hg.1 |
ENST00000595483.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010708.hg.1 |
ENST00000595483.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010708.hg.1 |
ENST00000595483.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010708.hg.1 |
ENST00000595483.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010708.hg.1 |
ENST00000595483.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010708.hg.1 |
ENST00000595483.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010708.hg.1 |
ENST00000595483.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010708.hg.1 |
ENST00000597870.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010708.hg.1 |
ENST00000597870.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010708.hg.1 |
ENST00000597870.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010708.hg.1 |
ENST00000597870.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010708.hg.1 |
ENST00000597870.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010708.hg.1 |
ENST00000597870.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010708.hg.1 |
ENST00000597870.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010708.hg.1 |
ENST00000597870.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010708.hg.1 |
ENST00000597870.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010708.hg.1 |
ENST00000601346.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010708.hg.1 |
ENST00000601346.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010708.hg.1 |
ENST00000601346.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010708.hg.1 |
ENST00000601346.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010708.hg.1 |
ENST00000601346.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010708.hg.1 |
ENST00000601346.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010708.hg.1 |
NM_000713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010708.hg.1 |
NM_000713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010708.hg.1 |
NM_000713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010708.hg.1 |
NM_000713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010708.hg.1 |
NM_000713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010708.hg.1 |
NM_000713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010708.hg.1 |
NM_000713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010708.hg.1 |
NM_000713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010708.hg.1 |
NM_000713.2 |
Refseq |