Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024021.3 | NP_076926.2 | 6 - 7 | 828 | 135 | |
NM_148975.2 | NP_683876.1 | 5 - 6 | 682 | 135 | ||
XM_017017909.1 | XP_016873398.1 | 5 - 6 | 695 | 135 | ||
GenBank mRNA | AB013102.1 | - | 5 - 6 | 523 | 135 | |
AB022821.1 | - | 5 - 6 | 547 | 135 | ||
AF068288.1 | - | 5 - 6 | 616 | 135 | ||
AF237912.1 | - | 5 - 6 | 633 | 135 | ||
AF350500.1 | - | 4 - 5 | 586 | 135 | ||
AK075081.1 | - | 6 - 7 | 713 | 135 | ||
AK292089.1 | - | 5 - 6 | 637 | 135 | ||
AK314033.1 | - | 5 - 6 | 525 | 135 | ||
BC020648.1 | - | 5 - 6 | 557 | 4720088 | 135 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |