Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INPP5K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130766.2 | NP_570122.1 | 2 - 3 | 664 | 94 | |
XM_005256686.1 | XP_005256743.1 | 2 - 3 | 511 | 94 | ||
XM_017024755.1 | XP_016880244.1 | 2 - 3 | 511 | 94 | ||
GenBank mRNA | AB036830.1 | - | 2 - 3 | 447 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Homo sapiens Probe spans exons |