Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05019731.hg.1 |
TC05001409.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05019731.hg.1 |
ENST00000231524.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05019731.hg.1 |
ENST00000381018.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05019731.hg.1 |
NM_001656.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05019731.hg.1 |
NM_033227.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500176230.hg.1 |
ENST00000231524.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500176230.hg.1 |
ENST00000381018.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500176230.hg.1 |
NM_001656.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500176230.hg.1 |
NM_033227.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500176230.hg.1 |
ENST00000231524.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500176230.hg.1 |
ENST00000381018.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500176230.hg.1 |
NM_001656.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500176230.hg.1 |
NM_033227.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500176230.hg.1 |
TC0500010930.hg.1 |
Clariom D TC |