Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010474.hg.1 |
TC02000668.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010474.hg.1 |
ENST00000283195.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010474.hg.1 |
ENST00000409491.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010474.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
ENST00000425282.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
ENST00000425282.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
ENST00000425282.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
ENST00000425282.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
ENST00000495506.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
ENST00000495506.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
ENST00000495506.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
ENST00000495506.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
ENST00000495924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
ENST00000495924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
ENST00000495924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
ENST00000629728.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
ENST00000629728.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200161535.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200161535.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200161535.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200161535.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200161535.hg.1 |
TC0200008807.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
ENST00000283195.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
ENST00000425282.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
ENST00000425282.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
ENST00000425282.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
ENST00000425282.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
ENST00000495506.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
ENST00000495506.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
ENST00000495506.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
ENST00000495506.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
ENST00000495924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
ENST00000495924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
ENST00000495924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
ENST00000629728.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
ENST00000629728.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008807.hg.1 |
NM_006267.4 |
Refseq |