Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
ENST00000314485.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
ENST00000314485.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
ENST00000314485.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
ENST00000314485.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
ENST00000314485.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
ENST00000314485.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
ENST00000314485.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
ENST00000314485.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
ENST00000314485.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
ENST00000314485.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
ENST00000314485.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
ENST00000368086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
ENST00000368086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
ENST00000368086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
ENST00000368086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
ENST00000368086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
ENST00000368086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
ENST00000368086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
ENST00000368086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
ENST00000368086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
ENST00000368086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
ENST00000368086.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
ENST00000448417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
ENST00000448417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
ENST00000448417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
ENST00000448417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
ENST00000448417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
ENST00000448417.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
ENST00000460351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
ENST00000460351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
ENST00000460351.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
ENST00000614243.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
ENST00000614243.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
ENST00000614243.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
ENST00000614243.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
ENST00000614243.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
ENST00000614243.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
ENST00000614243.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
ENST00000614243.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
ENST00000614243.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
ENST00000614243.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
NM_001206665.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
NM_001206665.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
NM_001206665.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
NM_001206665.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
NM_001206665.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
NM_001206665.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
NM_001206665.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
NM_001206665.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
NM_001206665.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
NM_001206665.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
NM_052868.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
NM_052868.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
NM_052868.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
NM_052868.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
NM_052868.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
NM_052868.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
NM_052868.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
NM_052868.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
NM_052868.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016122.hg.1 |
NM_052868.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053039.hg.1 |
TC01003397.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053039.hg.1 |
ENST00000314485.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053039.hg.1 |
ENST00000358475.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053039.hg.1 |
ENST00000368086.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053039.hg.1 |
NM_001206665.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053039.hg.1 |
NM_052868.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053040.hg.1 |
TC01003397.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053040.hg.1 |
ENST00000314485.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053040.hg.1 |
ENST00000358475.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053040.hg.1 |
ENST00000368086.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053040.hg.1 |
NM_001206665.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053040.hg.1 |
NM_052868.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100222437.hg.1 |
ENST00000314485.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100222437.hg.1 |
ENST00000368086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100222437.hg.1 |
ENST00000614243.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100222437.hg.1 |
NM_001206665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100222437.hg.1 |
NM_052868.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100222437.hg.1 |
ENST00000314485.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100222437.hg.1 |
ENST00000368086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100222437.hg.1 |
ENST00000614243.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100222437.hg.1 |
NM_001206665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100222437.hg.1 |
NM_052868.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100222437.hg.1 |
TC0100016122.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
ENST00000314485.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
ENST00000314485.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
ENST00000314485.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
ENST00000314485.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
ENST00000314485.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
ENST00000314485.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
ENST00000314485.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
ENST00000314485.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
ENST00000314485.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
ENST00000314485.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
ENST00000314485.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
ENST00000368086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
ENST00000368086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
ENST00000368086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
ENST00000368086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
ENST00000368086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
ENST00000368086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
ENST00000368086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
ENST00000368086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
ENST00000368086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
ENST00000368086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
ENST00000368086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
ENST00000448417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
ENST00000448417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
ENST00000448417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
ENST00000448417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
ENST00000448417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
ENST00000448417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
ENST00000460351.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
ENST00000460351.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
ENST00000460351.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
ENST00000614243.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
ENST00000614243.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
ENST00000614243.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
ENST00000614243.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
ENST00000614243.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
ENST00000614243.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
ENST00000614243.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
ENST00000614243.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
ENST00000614243.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
ENST00000614243.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
NM_001206665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
NM_001206665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
NM_001206665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
NM_001206665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
NM_001206665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
NM_001206665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
NM_001206665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
NM_001206665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
NM_001206665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
NM_001206665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
NM_052868.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
NM_052868.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
NM_052868.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
NM_052868.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
NM_052868.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
NM_052868.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
NM_052868.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
NM_052868.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
NM_052868.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016122.hg.1 |
NM_052868.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100222438.hg.1 |
ENST00000314485.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100222438.hg.1 |
ENST00000368086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100222438.hg.1 |
ENST00000448417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100222438.hg.1 |
ENST00000614243.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100222438.hg.1 |
NM_001206665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100222438.hg.1 |
NM_052868.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100222438.hg.1 |
ENST00000314485.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100222438.hg.1 |
ENST00000368086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100222438.hg.1 |
ENST00000448417.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100222438.hg.1 |
ENST00000614243.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100222438.hg.1 |
NM_001206665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100222438.hg.1 |
NM_052868.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100222438.hg.1 |
TC0100016122.hg.1 |
Clariom D TC |