Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19023372.hg.1 |
ENST00000335875.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19023372.hg.1 |
ENST00000454748.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19023372.hg.1 |
NM_014037.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19023372.hg.1 |
TC19001710.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000335875.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000335875.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000335875.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000335875.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000335875.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000335875.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000335875.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000335875.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000335875.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000335875.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000335875.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000335875.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000335875.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000335875.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000335875.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000335875.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000335875.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000335875.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000335875.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000335875.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000454748.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000454748.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000454748.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000454748.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000454748.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000454748.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000454748.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000454748.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000454748.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000454748.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000454748.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000454748.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000454748.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000454748.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000454748.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000454748.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000454748.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000454748.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000454748.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000594180.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000594180.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000594180.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000594180.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000594917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000594917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000594917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000594917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000597969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000597969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000597969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000597969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000597969.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000598221.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000598221.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000598221.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000598221.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000598221.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000598828.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000598828.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000598828.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000598828.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000598828.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000598828.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
ENST00000598828.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
NM_014037.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
NM_014037.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
NM_014037.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
NM_014037.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
NM_014037.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
NM_014037.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
NM_014037.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
NM_014037.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
NM_014037.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
NM_014037.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
NM_014037.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
NM_014037.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
NM_014037.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
NM_014037.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
NM_014037.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
NM_014037.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
NM_014037.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
NM_014037.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
NM_014037.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011132.hg.1 |
NM_014037.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900193165.hg.1 |
ENST00000335875.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193165.hg.1 |
ENST00000454748.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193165.hg.1 |
ENST00000594917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193165.hg.1 |
NM_014037.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900193165.hg.1 |
TC1900011132.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900193165.hg.1 |
ENST00000335875.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193165.hg.1 |
ENST00000454748.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193165.hg.1 |
ENST00000594917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193165.hg.1 |
NM_014037.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900193163.hg.1 |
ENST00000335875.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193163.hg.1 |
ENST00000454748.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193163.hg.1 |
ENST00000594917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193163.hg.1 |
ENST00000597969.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193163.hg.1 |
NM_014037.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900193163.hg.1 |
TC1900011132.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900193163.hg.1 |
ENST00000335875.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193163.hg.1 |
ENST00000454748.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193163.hg.1 |
ENST00000594917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193163.hg.1 |
ENST00000597969.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193163.hg.1 |
NM_014037.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000335875.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000335875.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000335875.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000335875.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000335875.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000335875.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000335875.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000335875.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000335875.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000335875.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000335875.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000335875.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000335875.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000335875.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000335875.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000335875.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000335875.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000335875.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000335875.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000335875.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000454748.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000454748.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000454748.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000454748.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000454748.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000454748.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000454748.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000454748.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000454748.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000454748.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000454748.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000454748.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000454748.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000454748.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000454748.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000454748.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000454748.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000454748.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000454748.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000594180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000594180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000594180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000594180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000594917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000594917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000594917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000594917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000597969.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000597969.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000597969.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000597969.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000597969.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000598221.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000598221.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000598221.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000598221.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000598221.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000598828.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000598828.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000598828.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000598828.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000598828.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000598828.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
ENST00000598828.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
NM_014037.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
NM_014037.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
NM_014037.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
NM_014037.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
NM_014037.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
NM_014037.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
NM_014037.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
NM_014037.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
NM_014037.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
NM_014037.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
NM_014037.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
NM_014037.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
NM_014037.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
NM_014037.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
NM_014037.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
NM_014037.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
NM_014037.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
NM_014037.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
NM_014037.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011132.hg.1 |
NM_014037.2 |
Refseq |