Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
ENST00000377705.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
ENST00000377705.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
ENST00000377705.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
ENST00000377705.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
ENST00000377705.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
ENST00000377705.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
ENST00000377705.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
ENST00000377705.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
ENST00000377705.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
ENST00000377705.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
ENST00000377705.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
ENST00000377705.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
ENST00000377705.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
ENST00000377705.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
ENST00000377705.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
ENST00000377705.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
ENST00000377705.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
ENST00000377705.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
ENST00000377705.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
ENST00000460777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
ENST00000460777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
ENST00000460777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
ENST00000460777.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
ENST00000462798.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
ENST00000462798.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
ENST00000462798.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
ENST00000464383.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
ENST00000464383.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
ENST00000464383.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
ENST00000464383.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
ENST00000464383.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
ENST00000464383.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
ENST00000464383.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
ENST00000464665.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
ENST00000464665.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
ENST00000464665.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
NM_024654.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
NM_024654.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
NM_024654.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
NM_024654.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
NM_024654.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
NM_024654.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
NM_024654.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
NM_024654.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
NM_024654.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
NM_024654.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
NM_024654.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
NM_024654.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
NM_024654.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
NM_024654.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
NM_024654.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
NM_024654.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
NM_024654.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
NM_024654.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
NM_024654.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012687.hg.1 |
NM_024654.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033268.hg.1 |
TC01002154.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033268.hg.1 |
ENST00000377705.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033268.hg.1 |
NM_024654.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100194027.hg.1 |
ENST00000377705.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100194027.hg.1 |
NM_024654.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100194027.hg.1 |
ENST00000377705.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100194027.hg.1 |
NM_024654.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100194027.hg.1 |
TC0100012687.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
ENST00000377705.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
ENST00000377705.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
ENST00000377705.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
ENST00000377705.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
ENST00000377705.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
ENST00000377705.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
ENST00000377705.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
ENST00000377705.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
ENST00000377705.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
ENST00000377705.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
ENST00000377705.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
ENST00000377705.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
ENST00000377705.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
ENST00000377705.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
ENST00000377705.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
ENST00000377705.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
ENST00000377705.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
ENST00000377705.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
ENST00000377705.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
ENST00000460777.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
ENST00000460777.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
ENST00000460777.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
ENST00000460777.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
ENST00000462798.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
ENST00000462798.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
ENST00000462798.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
ENST00000464383.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
ENST00000464383.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
ENST00000464383.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
ENST00000464383.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
ENST00000464383.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
ENST00000464383.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
ENST00000464383.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
ENST00000464665.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
ENST00000464665.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
ENST00000464665.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
NM_024654.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
NM_024654.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
NM_024654.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
NM_024654.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
NM_024654.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
NM_024654.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
NM_024654.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
NM_024654.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
NM_024654.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
NM_024654.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
NM_024654.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
NM_024654.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
NM_024654.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
NM_024654.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
NM_024654.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
NM_024654.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
NM_024654.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
NM_024654.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
NM_024654.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012687.hg.1 |
NM_024654.4 |
Refseq |