Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036642.hg.1 |
TC02002279.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036642.hg.1 |
ENST00000344908.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036642.hg.1 |
ENST00000409947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036642.hg.1 |
NM_006609.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
ENST00000344908.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
ENST00000344908.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
ENST00000344908.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
ENST00000344908.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
ENST00000344908.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
ENST00000344908.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
ENST00000344908.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
ENST00000344908.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
ENST00000344908.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
ENST00000344908.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
ENST00000344908.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
ENST00000344908.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
ENST00000344908.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
ENST00000344908.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
ENST00000344908.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
ENST00000344908.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
ENST00000344908.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
ENST00000409179.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
ENST00000409179.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
ENST00000409179.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
ENST00000409947.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
ENST00000409947.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
ENST00000409947.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
ENST00000409947.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
ENST00000409947.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
ENST00000409947.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
ENST00000409947.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
ENST00000409947.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
ENST00000409947.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
ENST00000409947.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
ENST00000409947.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
ENST00000409947.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
ENST00000409947.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
ENST00000409947.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
ENST00000409947.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
ENST00000409947.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
ENST00000409947.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
ENST00000409947.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
ENST00000409947.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
NM_006609.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
NM_006609.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
NM_006609.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
NM_006609.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
NM_006609.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
NM_006609.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
NM_006609.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
NM_006609.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
NM_006609.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
NM_006609.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
NM_006609.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
NM_006609.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
NM_006609.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
NM_006609.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
NM_006609.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
NM_006609.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
NM_006609.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
NM_006609.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
NM_006609.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200014170.hg.1 |
NM_006609.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200203495.hg.1 |
ENST00000344908.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203495.hg.1 |
ENST00000409947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203495.hg.1 |
NM_006609.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200203495.hg.1 |
ENST00000344908.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203495.hg.1 |
ENST00000409947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200203495.hg.1 |
NM_006609.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200203495.hg.1 |
TC0200014170.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
ENST00000344908.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
ENST00000344908.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
ENST00000344908.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
ENST00000344908.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
ENST00000344908.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
ENST00000344908.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
ENST00000344908.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
ENST00000344908.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
ENST00000344908.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
ENST00000344908.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
ENST00000344908.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
ENST00000344908.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
ENST00000344908.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
ENST00000344908.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
ENST00000344908.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
ENST00000344908.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
ENST00000344908.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
ENST00000409179.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
ENST00000409179.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
ENST00000409179.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
ENST00000409947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
ENST00000409947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
ENST00000409947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
ENST00000409947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
ENST00000409947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
ENST00000409947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
ENST00000409947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
ENST00000409947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
ENST00000409947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
ENST00000409947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
ENST00000409947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
ENST00000409947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
ENST00000409947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
ENST00000409947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
ENST00000409947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
ENST00000409947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
ENST00000409947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
ENST00000409947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
ENST00000409947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
NM_006609.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
NM_006609.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
NM_006609.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
NM_006609.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
NM_006609.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
NM_006609.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
NM_006609.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
NM_006609.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
NM_006609.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
NM_006609.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
NM_006609.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
NM_006609.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
NM_006609.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
NM_006609.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
NM_006609.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
NM_006609.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
NM_006609.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
NM_006609.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
NM_006609.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200014170.hg.1 |
NM_006609.4 |
Refseq |